Skip to Content
Merck
All Photos(1)

Key Documents

39391

Sigma-Aldrich

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide

≥97.0% (T), for peptide synthesis

Synonym(s):

N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide, EDC, WSC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N3
CAS Number:
Molecular Weight:
155.24
Beilstein:
507429
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Product Name

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)

Assay

≥97.0% (T)

form

liquid

reaction suitability

reaction type: Coupling Reactions

refractive index

n20/D 1.461

density

0.877 g/mL at 20 °C (lit.)

application(s)

peptide synthesis

functional group

amine

storage temp.

−20°C

SMILES string

CCN=C=NCCCN(C)C

InChI

1S/C8H17N3/c1-4-9-8-10-6-5-7-11(2)3/h4-7H2,1-3H3

InChI key

LMDZBCPBFSXMTL-UHFFFAOYSA-N

General description

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide is commonly used in combination with N-hydroxysuccinimide (NHS) in carbodiimide coupling reaction to activate carboxyl group for coupling with amines to form amides.

Application

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide has been used for the surface functionalization of graphene quantum dots (GQDs) employed as sensing probes in Janus micromotors to detect enterobacterial contamination. It has also been used for the activation of folic acid, prior to its conjugation on silica nanoparticles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1A - STOT RE 2 Oral

Target Organs

Stomach,large intestine,lymph node

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Liu et al.
Colloids and surfaces. B, Biointerfaces, 181, 143-148 (2019-05-28)
CTC detection has great potential to provide crucial clinical information for early cancer diagnosis, patient prognosis, personalized therapy, and cancer progress monitoring etc. It has been proved that the disease progress is associated with an increase in mesenchymal CTCs, while
Folic acid-conjugated organically modified silica nanoparticles for enhanced targeted delivery in cancer cells and tumor in vivo.
Yin F
Journal of Material Chemistry B: Materials for Biology and Medicine, 3(29), 6081-6093 (2015)
Ruogu Qi et al.
Nature communications, 8(1), 2166-2166 (2017-12-20)
Advanced-stage epithelial ovarian cancers are amongst the most difficult to treat tumors and have proven to be refractory to most cytotoxic, molecularly targeted, or immunotherapeutic approaches. Here, we report that nanoparticle-drug conjugates (NDCs) of monomethyl auristatin E (MMAE) significantly increase
Yumei Wang et al.
Materials science & engineering. C, Materials for biological applications, 103, 109737-109737 (2019-07-28)
In this study, twisted rod-like chiral mesoporous silicas (CMSs) with discriminating chiral characteristics (D/L) were designed and biomimetic synthesized by using L- and d-alanine derivatives as templates, and employed as poorly water-soluble chiral drug ibuprofen (IBU) carriers. The morphology and
Anthony C Y Yau et al.
Scientific reports, 7(1), 14998-14998 (2017-11-10)
Mineral oils are extensively used in our daily life, in food, cosmetics, biomedicine, vaccines and in different industrial applications. However, exposure to these mineral oils has been associated with immune adjuvant effects and the development of autoimmune diseases. Here we

Articles

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service