Skip to Content
Merck
All Photos(1)

Key Documents

20-106

Sigma-Aldrich

pNPP (p-Nitrophenyl Phosphate)

Synonym(s):

Nitrophenyl phosphate, p-Nitrophenyl phosphate, pNPP substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

form

solid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

shipped in

wet ice

Application

Research Category
All
Used to detect the phosphatase activity by pNPP hydrolysis.

Packaging

packed as 2 x 5 mg tablets

Quality

Routinely evaluated by using one of the following kits: PTP Assay Kit 1, PTP Assay Kit 2, Ser/Thr Phosphatase Assay Kit 1, or Alkaline/Acid Phosphatase Assay Kit

Physical form

Tablet

Storage and Stability

2 years at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of platelet activating factor (PAF) supplementation in semen extender on viability and ATP content of cryopreserved canine spermatozoa.
W Kordan,M Lecewicz,R Strzezek,A Dzieko?ska,L Fraser
Polish Journal of Veterinary Sciences null
Elif Duman et al.
ACS biomaterials science & engineering, 5(9), 4750-4763 (2019-09-09)
Hydroxyapatite (HAP) is the major biomineral of bone. Despite the large number of studies addressing HAP formation, a fundamental understanding of the critical roles of HAP-forming proteins in vitro is needed. Effects of two HAP-interacting proteins, osteocalcin (OCN) and osteopontin
Rhiannon R Penkert et al.
Obesity (Silver Spring, Md.), 28(9), 1631-1636 (2020-08-12)
Individuals with obesity suffer from an increased susceptibility to severe respiratory viral infections and respond poorly to vaccinations, making it imperative to identify interventions. Recent evidence suggesting that obesity leads to tissue-specific vitamin A deficiency led to an investigation of
Anna C E Shafe et al.
Therapeutic advances in gastroenterology, 4(6), 343-363 (2011-11-02)
Despite the high prevalence of constipation and its related public health implications, there is relatively little research available on the condition from large epidemiological studies. The aim of this study was to investigate the epidemiology of general practitioner (GP)-diagnosed constipation
Enzyme immunoassay ELISA and EMIT.
E Engvall
Methods in enzymology, 70(A), 419-439 (1980-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service