Skip to Content
Merck
All Photos(1)

Key Documents

N1891

Sigma-Aldrich

p-Nitrophenyl phosphate

SIGMAFAST, chromogenic, tablet

Synonym(s):

pNPP tablets

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

Product Name

SIGMAFAST p-Nitrophenyl phosphate Tablets, tablet, To prepare 5 mL

form

tablet

solubility

deionized water: soluble 5 mL

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Developed for use as a soluble substrate for the detection of alkaline phosphatase activity in Enzyme Immunoassays (EIA). pNPP is the substrate of choice in high sensitivity alkaline phosphatase-based EIA assays.One pNPP tablet and one Tris buffer tablet are dissolved in deionized water and provide a ready-to-use substrate. Each SIGMAFAST pNPP tablet set yields a solution containing 1.0 mg/ml pNPP, 0.2 M Tris buffer. EIA applications using pNPP may be read in timed assays or stopped with 3N NaOH for delayed readings. SIGMAFAST require only the addition of distilled or deionized water to prepare an active substrate solution. Tablets are individually packaged in foil packets.
SIGMA FAST p-Nitrophenyl phosphate Tablets has been used:
  • to quantify non-secreted alkaline phosphatase to monitor differentiation of osteoblast-like cells (SaoS-2)
  • to stain murine preosteoblast MC3T3-E1 cells in alkaline phosphatase assay
  • as a substrate for ExtrAvidin® alkaline phosphatase to measure hemoglobin-bound α-synuclein using enzyme-linked immunosorbent assay (ELISA)
  • as a substrate for alkaline phosphatase conjugated antibodies for detection in ELISA of abirin toxin

Reconstitution

Each tablet set dissolved in 5 mL deionized water yields a ready-to-use buffered solution containing pNPP.

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany
SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conall H Watson et al.
PLoS neglected tropical diseases, 11(7), e0005786-e0005786 (2017-07-21)
Fiji, an upper-middle income state in the Pacific Ocean, has experienced an increase in confirmed case notifications of enteric fever caused by Salmonella enterica serovar Typhi (S. Typhi). To characterize the epidemiology of typhoid exposure, we conducted a cross-sectional sero-epidemiological
Zimu Wu et al.
International journal of clinical and experimental pathology, 12(5), 1757-1763 (2020-01-15)
Post-stroke induction of alpha-synuclein (AS), a neuronal protein implicated in the pathogenesis of Parkinson's disease (PD), has been demonstrated to induce secondary brain damage after cerebral ischemia. Therefore, understanding the expression and pathogenic modifications of AS is clinically meaningful for
Ronit Rosenfeld et al.
Nature communications, 12(1), 944-944 (2021-02-13)
The coronavirus disease 2019 (COVID-19) pandemic, caused by the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), exhibits high levels of mortality and morbidity and has dramatic consequences on human life, sociality and global economy. Neutralizing antibodies constitute a highly promising
Ruklanthi de Alwis et al.
The Journal of infectious diseases, 219(2), 295-304 (2018-10-16)
Nontyphoidal Salmonella (NTS) organisms are a major cause of gastroenteritis and bacteremia, but little is known about maternally acquired immunity and natural exposure in infant populations residing in areas where NTS disease is highly endemic. We recruited 503 pregnant mothers
Deborah L Burnett et al.
Immunity, 54(12), 2908-2921 (2021-11-18)
Viral mutations are an emerging concern in reducing SARS-CoV-2 vaccination efficacy. Second-generation vaccines will need to elicit neutralizing antibodies against sites that are evolutionarily conserved across the sarbecovirus subgenus. Here, we immunized mice containing a human antibody repertoire with diverse

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Protocols

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service