Skip to Content
Merck

Skip To

MAK461

pNPP Phosphatase Assay Kit

Sufficient for 500 colorimetric tests

Synonym(s):

Phosphatase Assay Kit, p-Nitrophenyl Phosphate Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

input

protein

detection method

colorimetric

relevant disease(s)

cancer; hematological disorder; gastrointestinal diseases; neurological disorders; cardiovascular diseases

storage temp.

−20°C

General description

Para-nitrophenyl phosphate (pNPP) is a chromogenic substrate for most phosphatases such as alkaline phosphatases, acid phosphatases, protein tyrosine phosphatases and serine/threonine phosphatases. The reaction yields para-nitrophenol, which becomes an intense yellow soluble product under alkaline conditions and can be conveniently measured at A405 nm on a spectrophotometer.

Application

The kit is suitable for the quantitative determination of phosphatase activity of protein phosphatases.
The pNPP Phosphatase Assay Kit may be used for:
  • Metabolism Research
  • Cardiovascular Research
  • Neurodegenerative Research

Biochem/physiol Actions

The pNPP Phosphatase Assay kit involves adding a single reagent to the phosphatase and measuring the product formation on an absorbance reader. The assay can be conveniently performed in cuvettes, tubes or multi-well plates at either room temperature or 37 °C.

Features and Benefits

Simplified Process: Enjoy a hassle-free procedure that can be completed within 30 minutes without wash or reagent transfer steps, saving you time and effort.

Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N4645P7998N9389
Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

300

detection method

colorimetric

detection method

-

detection method

-

detection method

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

input

protein

input

-

input

-

input

-

relevant disease(s)

cancer; hematological disorder; gastrointestinal diseases; neurological disorders; cardiovascular diseases

relevant disease(s)

-

relevant disease(s)

-

relevant disease(s)

-


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

target_organs

Nervous system,Kidney,Liver,Skin

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
MAK461-1KT04065267335773

Questions

Reviews

No rating value

Active Filters