Skip to Content
Merck
All Photos(5)

Key Documents

07-645

Sigma-Aldrich

Anti-Sp1 Antibody

Upstate®, from rabbit

Synonym(s):

Sp1 transcription factor, specificity protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
electrophoretic mobility shift assay: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SP1(6667)

General description

Sp1 is a human transcription factor involved in gene expression in the early development of an organism. The protein is 785 amino acids long, with a molecular weight of 95 kDA. The SP1 transcription factor contains a zinc finger protein motif, by which it binds directly to DNA and enhances gene transcription. Sp1 has always been described as a ubiquitous transcription factor that is required for the constitutive and inducible expression of a variety of genes, such as in cell cycle or mammalian development.

Specificity

Expected to cross-react with rat.
Recognizes Sp1, Mr 105 kDa and 95 kDa.

Immunogen

Full length human Sp1 protein.

Application

Immunoprecipitation:
4 µg of this lot immunoprecipitated Sp1 from 500 µg of HeLa nuclear extract.
NOTE: A previous lot successfully immunoprecipitated the α-1 subunit using the Catch and Release v2.0 Reversible Immunoprecipitation System (Catalog # 17-500). See manual for more information and protocol: www.millipore.com

Chromatin Immunoprecipitation:
Reported by an independent laboratory.

Immunofluorescence:
Reported by an independent laboratory.

Electrophoretic Mobility Shift Assay (EMSA):
1 µL of a previous lot of this antibody has been reported by an independent laboratory to supershift DNA complexes containing recombinant Sp1 and endogenous Sp1 from HeLa cells in EMSA.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-Sp1 Antibody (Rabbit Polyclonal Antibody) validated in ChIP, EMSA, IF, IP, WB to detect Sp1 also known as Sp1 transcription factor, specificity protein 1.

Quality

Routinely evaluated by Western Blot on HeLa lysates.

Western Blot Analysis:
1:500 dilution of this lot detected SP1 on 10 μg of HeLa lysates.

Target description

Varies

Linkage

Replaces: MABE550

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in buffer containing 0.014 M phosphate buffer, pH 7.6, 0.175 M NaCl, 0.07% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion hotspots in AMACR promoter CpG island are cis-regulatory elements controlling the gene expression in the colon.
Zhang, X; Leav, I; Revelo, MP; Deka, R; Medvedovic, M; Jiang, Z; Ho, SM
PLoS Genetics null
Yong Wang et al.
Clinical epigenetics, 8, 62-62 (2016-05-28)
Ultraviolet-B (UVB) exposure attributes to the formation of age-related nuclear cataract (ARNC), which is mediated with DNA damage. DNA damage, an important factor for pathogenesis of ARNC, is induced by UVB, and is generally resolved by the nucleotide excision repair
DHT selectively reverses Smad3-mediated/TGF-beta-induced responses through transcriptional down-regulation of Smad3 in prostate epithelial cells.
Song, K; Wang, H; Krebs, TL; Wang, B; Kelley, TJ; Danielpour, D
Molecular Endocrinology null
Constitutive expression of the brg1 gene requires GC-boxes near to the transcriptional start site.
Itoh T, Miyake K, Yamaguchi T, Tsuge M, Kaneoka H, Iijima S
Journal of Biochemistry null
Haipeng Sun et al.
Molecular endocrinology (Baltimore, Md.), 22(9), 2076-2084 (2008-07-05)
Cyclooxygenase (COX) encodes a rate-limiting enzyme in the biosynthesis of prostanoids. Although COX-1 is constitutively expressed in many tissues, we found that glucocorticoids cause elevated expression of COX-1 gene in cardiomyocytes. Corticosterone (CT) at physiologically relevant doses (0.05-1 microm) induces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service