Skip to Content
Merck
All Photos(5)

Documents

MABE343

Sigma-Aldrich

Anti-Puromycin Antibody, clone 12D10

clone 12D10, from mouse

Synonym(s):

Anti-Puromycin, Clone 12D10 Anti-Puromycin, Puromycin Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12D10, monoclonal

species reactivity

human

species reactivity (predicted by homology)

all

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NPEPPS(9520)

General description

Puromycin is an aminonucleoside antibiotic, derived from the Streptomyces alboniger bacterium, that functions as a protein synthesis inhibitor that blocks translation through premature chain termination in the ribosome. Monoclonal antibodies to puromycin may be used with standard immunochemical methods to directly monitor translation, a method known as surface sensing of translation (SUnSET). Part of the molecule resembles the 3′ end of the aminoacylated tRNA, making it useful for protein translation analysis. Puromycin induces DNA fragmentation in thymocytes and in human HL-60 leukemia cells.

Specificity

Anti-Puromycin Antibody, clone 12D10 demonstrated to react with Human test sample, preincubated with Puromycin. Predicted to react with all species when test sample is incubated with Puromycin.

Immunogen

Puromycin from Streptomyces alboniger

Application

Anti-Puromycin antibody, clone 12D10, detects puromycin incorporated into protein. Monoclonal antibodies to puromycin may be used with standard immunochemical methods.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Western Blotting Analysis (Total Protein Staining): HEK293 cell lysates treated with Puromycin and Cyclohexamide, or Puromycin only were resolved using SDS-PAGE and transferred to a membrane. Proteins were visualized using Ponceau S staining.

Immunocytochemistry Analysis: A 1:10,000 dilution from a representative lot detected Puromycin-incorporated neosynthesized proteins in HeLa cells treated with Puromycin.

Western Blotting Analysis: A representative lot detected Puromycin-incorporated neosynthesized proteins in WB (Reineke, L. C., et al. (2012). Mol Biol Cell. 23(18):3499-3510.; Trinh, M. A. et al. (2012). Cell Rep. 1(6):678-688.; Fortin, D. A., et al. (2012). J Neurosci. 32(24):8127-8137.; Clavarino, G., et al. (2012). PLoS Pathog. 8(5):e1002708.; David, A., et al. (2012). J Cell Biol. 197(1):45-57.; White, L. K., et al. (2011). J Virol. 85(1):606-620.; Hoeffer, C. A., et al. (2011). Proc Natl Acad Sci USA. 108(8):3383-3388.; Goodman, C. A., et al. (2010). FASEB J. 25(3):1028-1039.; Schmidt, E., K., et al. (2009). Nat Methods. 6(4):275-277.; Santini, E., et al. (2013). Nature. 493(7432):411-415.; Quy. P. N., et al. (2013). J Biol Chem. 288(2):1125-1134.; Hulmi. J. J., et al. (2012). Am J Physiol Endocrinol Metab. 304(1):E41-50.; Bhattacharya, A., et al. (2012). Neuron. 76(2):325-337.; Hoeffer, C. A., et al. (2013). J Neurophysiol. 109(1):68-76.).

Immunofluorescence Analysis: A representative lot detected Puromycin-incorporated neosynthesized proteins in WB (Reineke, L. C., et al. (2012). Mol Biol Cell. 23(18):3499-3510.; Trinh, M. A. et al. (2012). Cell Rep. 1(6):678-688.; Fortin, D. A., et al. (2012). J Neurosci. 32(24):8127-8137.;David, A., et al. (2012). J Cell Biol. 197(1):45-57.; David, A., et al. (2011). J Biol Chem. 286(23):20688-20700.; White, L. K., et al. (2011). J Virol. 85(1):606-620.; Hoeffer, C. A., et al. (2011). Proc Natl Acad Sci USA. 108(8):3383-3388.; Schmidt, E., K., et al. (2009). Nat Methods. 6(4):275-277.; Goodman, C. A., et al. (2012). Proc Natl Acad Sci USA. 109(17):E989.; Santini, E., et al. (2013). Nature. 493(7432):411-415.; Quy. P. N., et al. (2013). J Biol Chem. 288(2):1125-1134.).

Immunohistochemistry Analysis: A representative lot detecte Puromycin-incorporated neosynthesized protein in IHC (Goodman, C. A., et al. (2010). FASEB J. 25(3):1028-1039.).

Fluorescence Activated Cell Sorting Analysis: A representative lot detected Puromycin-incorporated neosynthesized proteins in FACS (David, A., et al. (2012). J Cell Biol. 197(1):45-57.; Schmidt, E., K., et al. (2009). Nat Methods. 6(4):275-277.).

Alexa Fluor is a registered trademark of Life Technologies.

Quality

Evaluated by Western Blotting in HEK293 cell lysates treated with Puromycin and Cyclohexamide, or with Puromycin only.

Western Blotting Analysis: A 1:25,000 dilution of this antibody detected Puromycin-incorporated neosynthesized proteins in HEK293 cell lysates treated with Puromycin only. This antibody also detected small mounts of Puromycin-incorporated neosynthesized proteins in HEK293 cells treated with Puromycin and Cyclohexamide.

Target description

Puromycin is incorporated in neosynthesized proteins. In the presense of Puromycin only, this antibody detects Puromycin-incorporated neosynthesized proteins at multiple molecular weights. However, a weaker signal is observed in the co-presense of Cycloheximide, an inhibitor of protein biosynthesis in eukaryotic organisms.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysates treated with Puromycin and Cyclohexamide, or with Puromycin only.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel W D West et al.
The Journal of physiology, 594(2), 453-468 (2015-11-10)
Ribosome biogenesis is the primary determinant of translational capacity, but its regulation in skeletal muscle following acute resistance exercise is poorly understood. Resistance exercise increases muscle protein synthesis acutely, and muscle mass with training, but the role of translational capacity
Anna Maria Copeland et al.
Journal of virology, 87(21), 11659-11669 (2013-08-24)
Rift Valley fever virus (RVFV), an ambisense member of the family Bunyaviridae, genus Phlebovirus, is the causative agent of Rift Valley fever, an important zoonotic infection in Africa and the Middle East. Phlebovirus proteins are translated from virally transcribed mRNAs
Kim Tran et al.
ACS chemical biology, 9(12), 2796-2806 (2014-10-04)
Endoplasmic reticulum (ER) stress plays an important role in the decline in pancreatic β cell function and mass observed in type 2 diabetes. Here, we developed a novel β cell-based high-throughput screening assay to identify small molecules that protect β
Thomas Chaillou et al.
Journal of cellular physiology, 231(9), 1894-1902 (2015-12-20)
The ribosome has historically been considered to have no cell-specific function but rather serve in a "housekeeping" capacity. This view is being challenged by evidence showing that heterogeneity in the protein composition of the ribosome can lead to the functional
Hanna Hörnberg et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(50), 12697-12706 (2016-12-16)
The establishment of precise topographic maps during neural development is facilitated by the presorting of axons in the pathway before they reach their targets. In the vertebrate visual system, such topography is seen clearly in the optic tract (OT) and

Articles

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Protocols

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service