Skip to Content
Merck
All Photos(1)

Key Documents

SML2135

Sigma-Aldrich

Biotinyl tyramide

≥97% (HPLC), powder, tyramide signal amplifier

Synonym(s):

(3aS,4S,6aR)-Hexahydro-N-[2-(4-hydroxyphenyl)ethyl]-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, BP, Biotin-Phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25N3O3S
CAS Number:
Molecular Weight:
363.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Biotinyl tyramide, ≥97% (HPLC)

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NCCC1=CC=C(O)C=C1)CCCC[C@@H]2SC[C@@]3([H])[C@]2([H])NC(N3)=O

InChI

1S/C18H25N3O3S/c22-13-7-5-12(6-8-13)9-10-19-16(23)4-2-1-3-15-17-14(11-25-15)20-18(24)21-17/h5-8,14-15,17,22H,1-4,9-11H2,(H,19,23)(H2,20,21,24)/t14-,15-,17-/m0/s1

InChI key

VZWXNOBHWODXCW-ZOBUZTSGSA-N

Application

Biotinyl tyramide or Biotin-Phenol has been used in proximity-dependent labeling for protein identification.[1][2]

Biochem/physiol Actions

Biotinyl tyramide is a reagent used for tyramide signal amplification for both immunohistochemistry (IHC) and in situ hybridization protocols and with either chromogenic or fluorescence detection. Preliminary binding of a probe is followed by secondary detection of the probe with an HRP-labeled antibody or streptavidin conjugate. Catalysis by the HRP results in the deposition of multiple biotinyl tyramide molecules in the immediate vicinity of the probe that can then be detected with a labeled streptavidin conjugate. Detection sensitivity can be 100-fold or more sensitive compared to conventional detection procedures.
Reagent used for tyramide signal amplification

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nguyen Thi Trang et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 27(3), 326-331 (2015-04-10)
This study evaluated the sensitivity of biotinyl-tyramide-based in situ hybridization (TISH) method by comparison with chromogenic in situ hybridization (CISH) and immunohistochemical staining (IHC) methods. This study also determined the effect of fixative and fixation time on the detection of
J F Breininger et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 48(12), 1593-1599 (2000-12-02)
To increase the sensitivity of fluorescence in situ hybridization (FISH) for detection of low-abundance mRNAs, we performed FISH on cryostat sections of rat hypothalamus with biotin-labeled riboprobes to leptin receptor (ObRb) and amplified the signal by combining tyramide signal amplification
Xiangru Huo et al.
Molecular cell, 77(2), 368-383 (2019-11-05)
Interphase chromatin is hierarchically organized into higher-order architectures that are essential for gene functions, yet the biomolecules that regulate these 3D architectures remain poorly understood. Here, we show that scaffold attachment factor B (SAFB), a nuclear matrix (NM)-associated protein with
Marina Yu Mazina et al.
Scientific reports, 10(1), 4793-4793 (2020-03-18)
Proximity-dependent biotin labelling revealed undescribed participants of the ecdysone response in Drosophila. Two labelling enzymes (BioID2 and APEX2) were fused to EcR or Usp to biotin label the surrounding proteins. The EcR/Usp heterodimer was found to collaborate with nuclear pore
Misuzu Kurihara et al.
Molecular cell, 78(3), 493-505 (2020-05-01)
The promyelocytic leukemia (PML) body is a phase-separated nuclear structure physically associated with chromatin, implying its crucial roles in genome functions. However, its role in transcriptional regulation is largely unknown. We developed APEX-mediated chromatin labeling and purification (ALaP) to identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service