Skip to Content
Merck
All Photos(2)

Key Documents

850757C

Avanti

16:0-18:1 PE

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine; POPE; PE(16:0/18:1(9Z)); 110637

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76NO8P
CAS Number:
Molecular Weight:
718.00
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine, chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850757C-25mg)
pkg of 2 × 20 mL (850757C-1g)
pkg of 2 × 4 mL (850757C-200mg)
pkg of 5 × 4 mL (850757C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (850757C-25mg)
25 mg/mL (850757C-1g)
25 mg/mL (850757C-200mg)
25 mg/mL (850757C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H76NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,37H,3-16,19-36,40H2,1-2H3,(H,43,44)/b18-17-

InChI key

FHQVHHIBKUMWTI-ZCXUNETKSA-N

Application

16:0-18:1 PE has been used as a synthetic lipid for the preparation of liposomes.[1][2]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850757C-1g)
5 mL Clear Glass Sealed Ampule (850757C-200mg)
5 mL Clear Glass Sealed Ampule (850757C-25mg)
5 mL Clear Glass Sealed Ampule (850757C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondria: Practical Protocols (2018)
Ionization Properties of Phospholipids Determined by Zeta Potential Measurements
Sathappa M and Alder NN
Bio-protocol, 6(22) (2016)
Mariya Chavarha et al.
Langmuir : the ACS journal of surfaces and colloids, 28(48), 16596-16604 (2012-11-13)
Prior studies have shown that the biological mixture of the two hydrophobic surfactant proteins, SP-B and SP-C, produces faster adsorption of the surfactant lipids to an air/water interface, and that they induce 1-palmitoyl-2-oleoyl phosphatidylethanolamine (POPE) to form inverse bicontinuous cubic
Tânia Silva et al.
Langmuir : the ACS journal of surfaces and colloids, 34(5), 2158-2170 (2018-01-07)
An understanding of the mechanism of action of antimicrobial peptides is fundamental to the development of new and more active antibiotics. In the present work, we use a wide range of techniques (SANS, SAXD, DSC, ITC, CD, and confocal and
Sónia Troeira Henriques et al.
Journal of the American Chemical Society, 141(51), 20460-20469 (2019-11-26)
Peptides with pharmaceutical activities are attractive drug leads, and knowledge of their mode-of-action is essential for translation into the clinic. Comparison of native and enantiomeric peptides has long been used as a powerful approach to discriminate membrane- or receptor-mediated modes-of-action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service