Skip to Content
Merck
All Photos(1)

Key Documents

W268305

Sigma-Aldrich

Methyl benzoate

≥98%, FCC, FG

Synonym(s):

Bnzoic acid methyl ester, Clorius, Methyl benzene carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15
FEMA Number:
2683
Beilstein:
1072099
EC Number:
Council of Europe no.:
260
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.725
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

4.68 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.516 (lit.)

bp

198-199 °C (lit.)

mp

−12 °C (lit.)

solubility

ethanol: 60%, clear (1mL/4ml)

density

1.088 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; phenolic; wintergreen

SMILES string

COC(=O)c1ccccc1

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

QPJVMBTYPHYUOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Iqbal Choudhary et al.
Phytochemistry, 69(4), 1018-1023 (2008-01-08)
Two glycosides, 6'-O-(3,4-dihydroxy benzoyl)-beta-D-glucopyranosyl ester (1), and 4-O-beta-d-glucopyranoside-3-hydroxy methyl benzoate (2), along with five known compounds methyl benzoate (3), hypogallic acid (4), caffeic acid (5), paeoniflorin (6) and pikuroside (7) were isolated for the first time from a fresh water
Gianfranco Cainelli et al.
Chemistry & biodiversity, 5(5), 811-829 (2008-05-22)
Novel 4-alkylidene-beta-lactams with a polyphenolic side chain were synthesized and evaluated as radical scavengers. We have undertaken a detailed study of the antioxidant activity in vitro with chemical and biological testing of the new beta-lactams and of the corresponding methyl
Nan Zhao et al.
Plant physiology and biochemistry : PPB, 48(4), 279-287 (2010-03-05)
Two benzenoid esters, methyl salicylate (MeSA) and methyl benzoate (MeBA), were detected from insect-damaged rice plants. By correlating metabolite production with gene expression analysis, five candidate genes encoding putative carboxyl methyltransferases were identified. Enzymatic assays with Escherichia coli-expressed recombinant proteins
Derek R Boyd et al.
Organic & biomolecular chemistry, 8(6), 1415-1423 (2010-03-06)
The cis-dihydrodiol metabolite from methyl benzoate has been used as a synthetic precursor of carba-beta-L-galactopyranose, carba-beta-L-talopyranose and carba-alpha-L-talopyranose. The structures and absolute configurations of these carbasugars were determined by a combination of NMR spectroscopy, stereochemical correlation and X-ray crystallography.
N Sundaraganesan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(3), 771-777 (2007-04-17)
The Fourier transform Raman and Fourier transform infrared spectra of methyl benzoate (MB) were recorded in the liquid phase. The equilibrium geometry, harmonic vibrational frequencies, infrared intensities and Raman scattering activities, depolarization ratios, reduced masses were calculated by Hartree-Fock (HF)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service