Skip to Content
Merck
All Photos(1)

Key Documents

438073

Sigma-Aldrich

Nitric acid

Nitric acid

ACS reagent, 70%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38060304
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor pressure

8 mmHg ( 20 °C)

Assay

68.0-70.0% (ACS specification)
70%

form

liquid

ign. residue

≤5 ppm

color

APHA: ≤10

pH

<1.0

bp

120.5 °C (lit.)

density

1.413 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 ppm
sulfate (SO42-): ≤1 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
heavy metals (as Pb): ≤0.2 ppm

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitric acid (HNO3) is a highly corrosive mineral acid.[1][2] It is usually utilized for abstracting transition metal catalyst from single-walled carbon nanotubes (SWNTs) in order to purify them.[3] The effect of nitric acid on the oxidation of multiwalled carbon nanotubes (MWCNTs) has been investigated by sample weight, Raman spectrum, solubility, morphology and alignment examinations.[4]

Application

Nitric acid was used to increase the number of acid sites by altering the surface of activated carbon.[5]
It may be used:
  • In combination with acetic anhydride and zeolite catalysts for the preparation of p-nitrotoluene with high regioselectivity.[6]
  • In the pretreatment of rice straw for enhanced production of xylose.[7]
  • To minimize the extent of cation ordering in LiNi0.5Mn1.5O4, a transition metal-substituted spinel material, by surface treatment.[8]

Other Notes

SKU-Pack sizes with -P are packaged in poly bottles

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ilgook Kim et al.
Carbohydrate polymers, 99, 563-567 (2013-11-28)
In this study, nitric acid (HNO₃) was evaluated as an acid catalyst for rice straw pretreatment, and, after neutralization, as a sole nitrogen source for subsequent fermentation. Response surface methodology was used to obtain optimal pretreatment condition with respect to
Structurally stabilized LiNi0.5 Mn1.5O4 with enhanced electrochemical properties through nitric acid treatment.
Park JS, et al.
Journal of Power Sources, 230, 138-142 (2013)
Zeolite-Assisted Regioselective Nitration of Toluene in Acetonitrile with Nitric Acid/Acetic Anhydride.
Shi C and Tai Y.
Asian Journal of Chemistry, 27(7), 2420-2422 (2015)
Nitric acid purification of single-walled carbon nanotubes.
Hu H, et al.
The Journal of Physical Chemistry B, 107(50), 13838-13842 (2003)
THE TREATMENT OF ACID AND ALKALI BURNS: AN EXPERIMENTAL STUDY.
E C Davidson
Annals of surgery, 85(4), 481-489 (1927-04-01)

Articles

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1–2 of 3 Reviews  

Active Filters

  1. PA
    • Review 1
    • Votes 2
    5 out of 5 stars.

    excellent

    Fast delivery, good product. Was in need and they delivered

    Helpful?

    • Review 1
    • Votes 2
    5 out of 5 stars.

    Nitric acid

    This product arrived quickly and safely. Product is as promised.

    Helpful?

  2. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service