Skip to Content
Merck
All Photos(1)

Key Documents

319961

Sigma-Aldrich

Carbon tetrachloride

reagent grade, 99.9%

Synonym(s):

Tetrachloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl4
CAS Number:
Molecular Weight:
153.82
Beilstein:
1098295
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
Assay:
99.9%
bp:
76-77 °C (lit.)
vapor pressure:
143 mmHg ( 30 °C)
91 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

5.32 (vs air)

vapor pressure

143 mmHg ( 30 °C)
91 mmHg ( 20 °C)

Assay

99.9%

form

liquid

refractive index

n20/D 1.460 (lit.)

bp

76-77 °C (lit.)

mp

−23 °C (lit.)

density

1.594 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)Cl

InChI

1S/CCl4/c2-1(3,4)5

InChI key

VZGDMQKNWNREIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbon tetrachloride (CCl4) is a volatile and stable chlorinated hydrocarbon. It is widely used as a chemical intermediate, solvent, and dry-cleaning agent.

Application

Carbon tetrachloride can be used as:      
  • A precursor to synthesize high-surface-area mesoporous carbon material by a self-templating method in the presence of sodium-potassium alloy (NaK) as a reducing agent.      
  • A reactant to synthesize 1,2-disubstituted cyclopentanes via Cu-catalyzed ring-closing Kharasch addition reaction with 1,6-dienes.      
  • A solvent as well as a reactant to prepare diphenyl carbonate from phenol and carbon dioxide in the presence of ZnCl2 as a catalyst.

Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Ozone 1 - Skin Sens. 1B - STOT RE 1 Inhalation

Target Organs

Liver,Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seham S El-Din El-Hawary et al.
Antioxidants (Basel, Switzerland), 8(12) (2019-12-19)
The prevalence of hepatic diseases globally and in Egypt particularly necessitates an intensive search for natural hepatoprotective candidates. Despite the traditional use of Chrysophyllum oliviforme L. and C. cainito L. leaves in the treatment of certain ailments, evidence-based reports on
Ruihua Ma et al.
eLife, 9 (2020-03-11)
The distribution of complementary metabolic functions in hepatocytes along a portocentral axis is called liver zonation. Endothelial secreted Wnt ligands maintain metabolic zonation in the adult murine liver but whether those ligands are necessary to initiate zonation in the immature
Marc Mejias et al.
Gut, 64(4), 657-666 (2014-05-23)
Antiangiogenic strategies have been proposed as a promising new approach for the therapy of portal hypertension and chronic liver disease. Pigment epithelium-derived factor (PEDF) is a powerful endogenous angiogenesis inhibitor whose role in portal hypertension remains unknown. Therefore, we aimed
Meritxell Huch et al.
Nature, 494(7436), 247-250 (2013-01-29)
The Wnt target gene Lgr5 (leucine-rich-repeat-containing G-protein-coupled receptor 5) marks actively dividing stem cells in Wnt-driven, self-renewing tissues such as small intestine and colon, stomach and hair follicles. A three-dimensional culture system allows long-term clonal expansion of single Lgr5(+) stem
Bi-Sen Ding et al.
Nature, 505(7481), 97-102 (2013-11-22)
Chemical or traumatic damage to the liver is frequently associated with aberrant healing (fibrosis) that overrides liver regeneration. The mechanism by which hepatic niche cells differentially modulate regeneration and fibrosis during liver repair remains to be defined. Hepatic vascular niche

Questions

1–5 of 5 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Who do I contact about larger solvent volume needs?

    1 answer
    1. Email us at [email protected] or visit Sigma-Aldrich Fine Chemicals for development and manufacturing-scale inquiries: [email protected]

      Helpful?

  3. What apparatus do you recommend for HPLC solvent filtration?

    1 answer
    1. The Sigma-Aldrich Vacuum Filtration Assembly gives you all the components you need including: funnel top, fritted glass funnel support, filtration flask, aluminum clamp, and silicone stopper.  The Acrodisc Syringe Filters offer high quality filtration for analytical samples, certified for HPLC to ensure low extractable and available in a broad range of membranes to meet sample compatibility requirements.

      Helpful?

  4. Can you recommend a Solvent Extractor for organics from aqueous solution?

    1 answer
    1. Solvent extractors are often used for the quantitative extraction of organics from aqueous solutions and separations in immunoassays. The MIXXOR system has been applied successfully in many laboratory solvent extraction operations and is ideal for the rapid screening of alternative solvents for specific extraction problems.

      Helpful?

  5. What are the options for drying solvents?

    1 answer
    1. Sigma-Aldrich offers a full range of high-purity solvents with extremely low water levels specifically manufactured for moisture sensitive Organic and Biotech applications.  Sigma-Aldrich also carries various drying agents such as molecular sieves, which are typically compatible with organic solvents.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service