Skip to Content
Merck
All Photos(3)

Key Documents

W262701

Sigma-Aldrich

Levulinic acid

greener alternative

natural, 99%, FG

Synonym(s):

4-Oxopentanoic acid, 4-Oxovaleric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COOH
CAS Number:
Molecular Weight:
116.12
FEMA Number:
2627
Beilstein:
506796
EC Number:
Council of Europe no.:
23
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.023
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

vapor pressure

1 mmHg ( 102 °C)

Assay

99%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.442

bp

245-246 °C (lit.)

mp

30-33 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

caramel; creamy; acidic; sweet; vanilla

SMILES string

CC(=O)CCC(O)=O

InChI

1S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)

InChI key

JOOXCMJARBKPKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Levulinic acid, an organic acid, is mainly used as a cigarette additive.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Use of carboxymethyl cellulose as binder for the production of water-soluble catalysts.: Levulinic acid is used in conjunction with carboxymethyl cellulose to produce water-soluble catalysts. This application demonstrates the acid′s versatility in catalyst synthesis, enhancing the solubility and functionality of the resulting products (Paganelli et al., 2024).

  • Nanocavity in hollow sandwiched catalysts as substrate regulator for boosting hydrodeoxygenation of biomass-derived carbonyl compounds.: This research explores the use of levulinic acid in biomass conversion processes. The study highlights the acid′s role in enhancing hydrodeoxygenation reactions, contributing to more efficient biofuel production (Zheng et al., 2024).

  • Mechanism of CO(2) in promoting the hydrogenation of levulinic acid to γ-valerolactone catalyzed by RuCl(3) in aqueous solution.: This paper investigates the catalytic hydrogenation of levulinic acid to γ-valerolactone. The findings provide insights into the role of CO2 in enhancing reaction efficiency, offering valuable information for industrial applications (Min et al., 2024).

  • Integrated biorefinery approach for utilization of wood waste into levulinic acid and 2-Phenylethanol production under mild treatment conditions.: This study presents a biorefinery approach to convert wood waste into valuable chemicals, including levulinic acid. The process demonstrates the potential for sustainable production of levulinic acid and its derivatives from renewable resources (Pachapur et al., 2024).


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suheyla Kocaman
International journal of phytoremediation, 22(8), 885-895 (2020-03-11)
This study has developed an innovative and environmentally friendly approach for the removal of methylene blue (MB) dye by natural shells (NShs) chemically modified with levulinic acid (LA). Almond shell (ASh), walnut shell (WSh), and apricot kernel shell (AKSh) were
Sofia Tallarico et al.
Scientific reports, 9(1), 18858-18858 (2019-12-13)
Cellulose is the main component of lignocellulosic biomass. Its direct chemocatalytic conversion into lactic acid (LA), a powerful biobased chemical platform, represents an important, and more easily scalable alternative to the fermentative way. In this paper, we present the selective
Conversion of hemicellulose to furfural and levulinic acid using biphasic reactors with alkylphenol solvents.
Elif I Gürbüz et al.
ChemSusChem, 5(2), 383-387 (2012-01-26)
Richard D Ashby et al.
Bioresource technology, 118, 272-280 (2012-06-19)
Glycerine (a biodiesel co-product) and levulinic acid (a pulp and paper co-product) were used as co-substrates for the fermentative synthesis of short-chain polyhydroxyalkanoate (sc-PHA) biopolymers with tunable monomer and molecular weight characteristics. Pseudomonas oleovorans NRRL B-14682 utilized glycerine alone to
Wenyang Pan et al.
Journal of industrial microbiology & biotechnology, 39(3), 459-469 (2011-09-29)
Sugar maple hemicellulosic hydrolysate containing 71.9 g/l of xylose was used as an inexpensive feedstock to produce polyhydroxyalkanoates (PHAs) by Burkholderia cepacia ATCC 17759. Several inhibitory compounds present in wood hydrolysate were analyzed for effects on cell growth and PHA production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service