Skip to Content
Merck
All Photos(4)

Key Documents

A7007

Sigma-Aldrich

S-(5′-Adenosyl)-L-methionine chloride dihydrochloride

≥75%

Synonym(s):

SAM, Active methionine, AdoMet, SAM chloride dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23ClN6O5S · 2 HCl
CAS Number:
Molecular Weight:
507.82
Beilstein:
6560002
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

synthetic (chemical)

Quality Level

Assay

≥75%

form

powder

color

white to off-white

solubility

H2O: 100 mg/mL

application(s)

cell analysis

shipped in

dry ice

storage temp.

−20°C

SMILES string

Cl.Cl.[Cl-].C[S+](CC[C@H](N)C(O)=O)C[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23

InChI

1S/C15H22N6O5S.3ClH/c1-27(3-2-7(16)15(24)25)4-8-10(22)11(23)14(26-8)21-6-20-9-12(17)18-5-19-13(9)21;;;/h5-8,10-11,14,22-23H,2-4,16H2,1H3,(H2-,17,18,19,24,25);3*1H/t7-,8+,10+,11+,14+,27?;;;/m0.../s1

InChI key

KBAFOJZCBYWKPU-XQVUROGGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S-(5′-Adenosyl)-L-methionine chloride dihydrochloride (SAM) facilitates the transfer of methyl groups to proteins, lipids and nucleic acids. Methionine adenosyl transferase catalyses the synthesis of SAM from methionine and adenosine triphosphate (ATP). SAM functions to regulate various cellular functions like cell division, cell death, transcription, genetic stability, oxidant/antioxidant balance and polyamine homeostasis. Therapeutically, SAM finds its application as a nutritional supplement in humans for various diseases like osteoarthritis and liver injury. SAM also regulates transsulfuration reactions by binding to the regulatory domain of key enzyme cystathionine-β-synthase (CBS).[1]

Application

S-(5′-Adenosyl)-L-methionine chloride dihydrochloride has been used as a medium supplement for S30 cell extract.[2] It also has been used for measuring the interaction with catechol-O-methyltransferase (COMT) by quartz crystal microbalance (QCM), surface plasmon resonance (SPR) and isothermal titration calorimetry (ITC) with supported lipid bilayers and vesicles.[3]

Biochem/physiol Actions

Methyl donor; cofactor for enzyme-catalyzed methylations, including catechol O-methyltransferase (COMT) and DNA methyltransferases (DNMT).
Methyl donor; cofactor for enzyme-catalyzed methylations, including catechol O-methyltransferase (COMT) and DNA methyltransferases (DNMT). Although present in all cells, it is concentrated in liver where 85% of all methylation reactions occur. It is also involved in regulating liver function, growth, and response to injury.

Caution

This material is 80-90% pure when prepared, but is very unstable. As much as 10% purity loss per day at 25 °C has been noted.

Analysis Note

Purity based on UV and HPLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andria V Rodrigues et al.
Chembiochem : a European journal of chemical biology, 21(5), 663-671 (2019-09-13)
We recently reported the discovery of phenylacetate decarboxylase (PhdB), representing one of only ten glycyl-radical-enzyme reaction types known, and a promising biotechnological tool for first-time biochemical synthesis of toluene from renewable resources. Here, we used experimental and computational data to
Ted M Lakowski et al.
Analytical biochemistry, 396(1), 158-160 (2009-09-08)
S-Adenosyl-L-homocysteine (AdoHcy) background signal in reactions with protein arginine N-methyltransferase 1 is investigated using an ultrahigh-performance liquid chromatography tandem mass spectrometry assay that measures AdoHcy. We identify three sources of AdoHcy background: enzymatic automethylation, AdoHcy contamination in commercial S-adenosyl-L-methionine (AdoMet)
Chemical communication between bacteria and cell-free gene expression systems within linear chains of emulsion droplets
Schwarz-Schilling M, et al.
Integrative Biology : Quantitative Biosciences from Nano to Macro, 8(4), 564-570 (2016)
Yan Zhang et al.
Archives of microbiology, 191(10), 773-783 (2009-09-05)
Enolase-phosphatase (E1), as an enzyme, is involved in methionine salvage pathway in many prokaryotic and eukaryotic organisms. But the identity and function of E1 in Xanthomonas oryzae pv. oryzae (Xoo) remain undetermined. Here, we report the cloning and characterization of
Membrane bound COMT isoform is an interfacial enzyme: general mechanism and new drug design paradigm
Magarkar A, et al.
Chemical Communications (Cambridge, England), 54(28), 3440-3443 (2018)

Articles

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service