Skip to Content
Merck
All Photos(4)

Key Documents

A3785

Sigma-Aldrich

5-Aminolevulinic acid hydrochloride

≥98%, suitable for ligand binding assays

Synonym(s):

δ-Aminolevulinic acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, 5-Aminolaevulinic acid hydrochloride, ALA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2C(O)CH2CH2COOH · HCl
CAS Number:
Molecular Weight:
167.59
Beilstein:
3690651
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

5-Aminolevulinic acid hydrochloride, ≥98%

Quality Level

Assay

≥98%

form

powder

technique(s)

ligand binding assay: suitable

mp

150 °C

storage temp.

−20°C

SMILES string

Cl[H].NCC(=O)CCC(O)=O

InChI

1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H

InChI key

ZLHFONARZHCSET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Aminolevulinic acid hydrochloride has been used to activate or inhibit heme biosynthesis in HeLa and K562 cell lines. It has also been used in photodynamic therapy for A431 tumor bearing mice.

Biochem/physiol Actions

5-Aminolevulinic acid (5-ALA) is an intermediate in heme biosynthesis and is useful in cancer treatment. It is a non-protein amino acid. 5-ALA also has applications in the field of agriculture. It is being studied as an inducing reagent for protoporphyrin IX (PPIX) dependent fluorescence diagnosis of metastatic lymph nodes. 5-ALA is used for photodynamic therapy of diseases, such as Paget′s disease and HPV infection-associated cervical condylomata acuminata.
Intermediate in heme biosynthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Bertino et al.
Pain, 160(12), 2766-2775 (2019-08-14)
Hereditary sensory and autonomic neuropathies (HSANs) are a group of clinically and genetically heterogeneous disorders of the peripheral nervous system mainly characterized by impaired nociception and autonomic dysfunction. We previously identified heme metabolism as a novel pathway contributing to sensory
Optimization of the heme biosynthesis pathway for the production of 5-aminolevulinic acid in Escherichia coli
Junli Zhang
Scientific Reports, 5 (2015)
Treatment of HPV infection-associated cervical condylomata acuminata with 5-aminolevulinic acid-mediated photodynamic therapy.
Wang HW
Photochemistry and Photobiology, 88(3), 565-569 (2012)
ALA-PpIX variability quantitatively imaged in A431
epidermoid tumors using in vivo ultrasoundfluorescence
tomography and ex vivo assay
Alisha V DSouza
SPIE Proc., 8931 null
The mitochondrial heme exporter FLVCR1b mediates erythroid differentiation
Deborah Chiabrando, et.al
The Journal of Clinical Investigation, 122(12), 4569-4579 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service