Skip to Content
Merck
All Photos(1)

Documents

W212709

Sigma-Aldrich

Benzaldehyde

≥98%, FG, FCC

Synonym(s):

Bitter almond

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHO
CAS Number:
Molecular Weight:
106.12
FEMA Number:
2127
Beilstein:
471223
EC Number:
Council of Europe no.:
101
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.013
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor density

3.65 (vs air)
3.7 (vs air)

vapor pressure

4 mmHg ( 45 °C)

Assay

≥98%

form

liquid

autoignition temp.

374 °F

expl. lim.

1.4 %, 20 °F

refractive index

n20/D 1.545 (lit.)

pH

5.9 (20 °C)

bp

178-179 °C (lit.)

mp

−26 °C (lit.)

density

1.044 g/cm3 at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

almond; cherry; sweet

SMILES string

O=Cc1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H

InChI key

HUMNYLRZRPPJDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzaldehyde is an aromatic flavor compound with a characteristic bitter almond taste and odor. It has been identified in the volatile fractions of mushroom, medlar seeds and cheese.

Application


  • Volatolomics-assisted characterization of the key odorants in green off-flavor black tea and their dynamic changes during processing.: This study highlights the role of Benzaldehyde in identifying and characterizing key odorants in green off-flavor black tea, using advanced volatolomics techniques. This research provides essential insights into flavor science and the impact of processing methods on tea quality (Yang et al., 2024).

  • Volatilome and flavor analyses based on e-nose combined with HS-GC-MS provide new insights into ploidy germplasm diversity in Platostoma palustre.: This research uses Benzaldehyde as a standard in electronic nose and gas chromatography-mass spectrometry analyses to explore plant volatilomes, significantly contributing to our understanding of plant biochemistry and genetic diversity (Zhong et al., 2024).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenaroli G
Fenaroli's Handbook of Flavor Ingredients, 40-40 (1971)
Conversion of phenylalanine to benzaldehyde initiated by an aminotransferase in Lactobacillus plantarum
Groot MNN & de Bont JA.
Applied and Environmental Microbiology, 64(8), 3009-3013 (1998)
Supercritical fluid extraction of volatile components from Bunium persicum Boiss.(black cumin) and Mespilus germanica L.(medlar) seeds
Pourmortazavi SM, et al.
J. Food Compos. Anal., 18(5), 439-446 (2005)
Volatile components of mushroom (Agaricus subrufecens)
Chen CC & Wu CM
Journal of Food Science, 49(4), 1208-1209 (1984)
Tom Craig et al.
Biotechnology and bioengineering, 110(4), 1098-1105 (2012-11-06)
Benzaldehyde, with its apricot and almond-like aroma, is the second most abundantly used molecule in the flavor industry, and is most commonly produced via chemical routes, such as by the oxidation of toluene. Biologically produced benzaldehyde, whether by extraction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service