Skip to Content
Merck
All Photos(1)

Key Documents

W213705

Sigma-Aldrich

Benzyl alcohol

≥99%, FCC, FG

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
FEMA Number:
2137
Beilstein:
878307
EC Number:
Council of Europe no.:
58
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.010
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515
FDA 21 CFR 175.105

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥99%

form

liquid

autoignition temp.

817 °F

shelf life

5 yr

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsamic; floral; fruity; sweet

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzyl alcohol is an aromatic alcohol that is used as a flavoring agent and fragrance ingredient.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Burdock GA
Encyclopedia of Food and Color Additives, 1, 261-262 (1997)
Fragrance material review on anisyl alcohol
Scognamiglio J, et al.
Food And Chemical Toxicology, 50, S140-S160 (2012)
Béatrice Hechler et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 232-243 (2005-03-29)
Our aim was to determine whether the newly described P2X1 antagonist NF449 [4,4',4'',4'''-(carbonylbis(imino-5,1,3-benzenetriylbis(carbonylimino)))tetrakis-benzene-1,3-disulfonic acid octasodium salt] could selectively antagonize the platelet P2X1 receptor and how it affected platelet function. NF449 inhibited alpha,beta-methyleneadenosine 5'-triphosphate-induced shape change (IC50 = 83 +/- 13
C L Froebe et al.
Dermatologica, 181(4), 277-283 (1990-01-01)
The relationship between the in vivo irritation potential of sodium lauryl sulfate (SLS) and linear alkyl benzene sulfonate (LAS) and the ability of these two surfactants to remove lipid from the stratum corneum (SC) in vitro were investigated. Either surfactant
A P De Leenheer et al.
Clinical chemistry, 31(1), 142-146 (1985-01-01)
For this sensitive RIA for 1 alpha,25-dihydroxyvitamin D, we used antibodies to 1 alpha,25-dihydroxycholecalciferol-3-hemisuccinate conjugated to bovine serum albumin, raised in rabbits. These antibodies show a high affinity for 1 alpha,25-dihydroxyvitamin D3 but cross react with other vitamin D metabolites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service