Skip to Content
Merck
All Photos(1)

Documents

W231118

Sigma-Aldrich

Citronellyl acetate

≥95%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
2311
EC Number:
Council of Europe no.:
202
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.012
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.445 (lit.)

bp

240 °C (lit.)

density

0.891 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

citronellol, geraniol

Organoleptic

green; citrus; fruity; floral; rose

SMILES string

CC(CCOC(C)=O)CC\C=C(\C)C

InChI

1S/C12H22O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,11H,5,7-9H2,1-4H3

InChI key

JOZKFWLRHCDGJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyang-Sook Choi
Journal of agricultural and food chemistry, 53(5), 1642-1647 (2005-03-03)
This study was conducted to determine the composition of kumquat (Fortunella japonica Swingle) cold-pressed peel oil and to determine which volatile components are primarily responsible for the aroma of this oil. Eighty-two compounds were identified in the oil by GC
S Oda et al.
Bioscience, biotechnology, and biochemistry, 65(8), 1917-1919 (2001-10-02)
A double coupling system, which couples metabolism of glucose and transacetylation, is a unique procedure for the production of acetic esters. In the novel coupling system described in this article, acetyl coenzyme A (acetyl-CoA) was supplied via metabolism of both
Jun Han et al.
Pest management science, 67(12), 1583-1588 (2011-06-16)
This study was aimed at assessing the fumigant toxicity of 14 essential oil constituents from lemon eucalyptus, Eucalyptus citriodora Hook, and another ten known compounds to females of acaricide-susceptible, chlorfenapyr-resistant, fenpropathrin-resistant, pyridaben-resistant and abamectin-resistant strains of Tetranychus urticae Koch. Menthol
Yuhong Tian et al.
Se pu = Chinese journal of chromatography, 23(6), 651-654 (2006-02-28)
The volatile constituents in leaves of Eucalyptus citriodora, including oil fraction and water-soluble fraction, were extracted and determined. Oil fraction of volatile components was obtained through steam distillation. Ether was used as the solvent to extract the water-soluble fraction of
Karin Förster-Fromme et al.
Microbiology (Reading, England), 154(Pt 3), 789-796 (2008-03-04)
The atuRABCDEFGH gene cluster is essential for acyclic terpene utilization (Atu) in Pseudomonas aeruginosa. The biochemical functions of most Atu proteins have not been experimentally verified; exceptions are AtuC/AtuF, which constitute the two subunits of geranyl-CoA carboxylase, the key enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service