Skip to Content
Merck
All Photos(1)

Key Documents

W212806

Sigma-Aldrich

Benzaldehyde dimethyl acetal

≥98%, FG

Synonym(s):

α,α-Dimethoxytoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)2
CAS Number:
Molecular Weight:
152.19
FEMA Number:
2128
Beilstein:
2044501
EC Number:
Council of Europe no.:
37
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.003
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.493 (lit.)

bp

87-89 °C/18 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; floral; fruity; wine-like

SMILES string

COC(OC)c1ccccc1

InChI

1S/C9H12O2/c1-10-9(11-2)8-6-4-3-5-7-8/h3-7,9H,1-2H3

InChI key

HEVMDQBCAHEHDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Sakairi et al.
Carbohydrate research, 246, 61-73 (1993-08-17)
Treatment of phenyl alpha-maltoside with an excess of alpha, alpha-dimethoxytoluene in the presence of (+)-10-camphorsulfonic acid, followed by partial hydrolysis to remove unstable acyclic acetal substituents, gave phenyl 3,2':4',6'-di-O-benzylidene-alpha-maltoside. Thus, one of the benzylidene groups formed an eight-membered cyclic acetal
Simona Matrella et al.
Macromolecular bioscience, 15(7), 927-940 (2015-03-18)
Intrinsic antimicrobial thermoplastic A(BC)n copolymers (n = 1, 2, 4), where A was poly(ethylene glycol) (PEG), BC was a random chain of methylmethacrylate (MMA), and alkyl-aminoethyl methacrylate (AAEMA), were synthesized and the antimicrobial activity and hemolyticity were evaluated on plaques obtained by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service