Skip to Content
Merck
All Photos(1)

Key Documents

W210901

Sigma-Aldrich

L-Ascorbic acid

L-Ascorbic acid

FCC, FG

Synonym(s):

L-AA, L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
FEMA Number:
2109
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
FG
Fragrance grade
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

grade

FG
Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.3013
FDA 21 CFR 182.8013

Assay

99.0-100.5% (iodometric)

form

crystalline
powder or crystals

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid, an organic acid[1], is found abundantly in citrus fruits.[2] It is generally used as an antioxidant and pH adjuster in the cosmetic industry. L-AA is also used as a preservative and nutrient supplement in foods.[3]

Application


  • Integrated metabolomics of big six Escherichia coli on pea sprouts to organic acid treatments.: Explores the antimicrobial effects of organic acids including L-Ascorbic acid on E. coli strains, offering insights into its use in food safety practices (Wang Y et al., 2022).

  • Molecular Scalpel to Chemically Cleave Metal-Organic Frameworks for Induced Phase Transition.: Although not directly related to typical biochemical applications of L-Ascorbic acid, this innovative research uses it in material science to influence molecular structures, which could inform nanotechnology applications in biochemistry (Zhou X et al., 2021).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Final report of the safety assessment of L-Ascorbic Acid, Calcium Ascorbate, Magnesium Ascorbate, Magnesium Ascorbyl Phosphate, Sodium Ascorbate, and Sodium Ascorbyl Phosphate as used in cosmetics.
Elmore AR.
International Journal of Toxicology, 24, 51-111 (2005)
Differential transcriptional regulation of L-ascorbic acid content in peel and pulp of citrus fruits during development and maturation
Alos E, et al.
Planta, 239(5), 1113-1128 (2014)
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 113-113 (2012)
Determination of l-ascorbic acid in wines by direct injection liquid chromatography using a polymeric column
Lopes P, et al.
Analytica Chimica Acta, 555(2), 242-245 (2006)
A rapid method for the determination of total L-ascorbic acid in fruits and vegetables by micellar electrokinetic capillary chromatography
Thompson CO and Trenerry VC
Food Chemistry, 53(1), 43-50 (1995)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. 혹시 200mg만 구매하는 것은 불가능 한가요? 그리고 성상시험이나 순도시험같은 시험을 진행할때 필요한 아스코르브산 표준품은 이 제품을 구매하는 것이 맞나요?

    1 answer
    1. This product is not intended for use as an analytical standard. Please see the link below to review several analytical standard product options which also offer smaller package sizes:
      https://www.sigmaaldrich.com/product/compare?term=50-81-7&focus=products&page=1&perpage=30&sort=relevance&type=cas_number&products=PHR1008%3ASIAL%3Asial%2CA1300000%3ASIAL%3Asial%2C47863%3ASUPELCO%3Asial%2C57803%3ASIAL%3Asial&returnUrl=%2Fsearch%2F50-81-7

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service