Skip to Content
Merck
All Photos(1)

Key Documents

1043003

USP

Ascorbic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

L-Ascorbic acid, L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ascorbic acid

manufacturer/tradename

USP

technique(s)

HPLC: suitable

mp

190-194 °C (dec.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ascorbic acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ascorbic Acid Tablets
  • Ascorbic Acid Injection
  • Ascorbic Acid Chewable Gels
  • Ascorbic Acid Compounded Oral Solution
  • Sodium Ascorbate
  • Calcium Ascorbate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

This product is part of USP Biologics Program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glutathione
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(3), 5054-5054 (2020)
Ascorbic Acid Chewable Gels
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(4), 4765-4765 (2019)
W Lee et al.
Clinical chemistry, 28(10), 2165-2169 (1982-10-01)
Measurements of ascorbic acid concentration in leukocytes by "high-performance" liquid chromatography (HPLC) provides better nutritional assessment, leading to better management, particularly of presymptomatic and critically ill patients. This procedure includes a simple, reproducible cell-separation technique that requires no more than
Ascorbic Acid Compounded Oral Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(4), 2626-2626 (2020)
Ascorbic Acid Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(6), 383-383 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service