Skip to Content
Merck
All Photos(4)

Key Documents

A0278

Sigma-Aldrich

L-Ascorbic acid

reagent grade

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

reagent grade

Assay

≥98% (with Iodine, titration)

form

powder

technique(s)

HPLC: suitable

color

white to light yellow

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 50 mg/mL

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

Ascorbic Acid, also known as Vitamin C, is a six-carbon lactone produced by plants and some animal species but not by humans and other primates.[1] It is a water-soluble vitamin.[2]

Application

L-Ascorbic acid has been used as a standard in high performance liquid chromatography (HPLC) for quantification of L-ascorbic acid.[3]
L-Ascorbic acid may be used as standard in assay for erythrocyte hemolysis mediated by peroxyl free radicals.[4] It may be used for the reduction of graphene oxide sheets.[5]

Biochem/physiol Actions

L-Ascorbic acid is an anti-oxidant. L-Ascorbic acid (vitamin C) is important for many enzymatic reactions for maintaining prosthetic metal ions in their reduced forms (for example, Fe2+, Cu+). It protects tissues from oxidative damage by scavenging free radicals.[6] It has mild reductive ability and nontoxic property. It is commonly employed reductant in the laboratory.[5] Colorimetric determination of ascorbic acid in plasma using acid phosphotungstate has been reported.[7]
L-ascorbic acid modified poly(ester urethane)s might facilitate soft tissue regeneration.[8] It also helps in increasing iron absorption by tissues. In addition, it aids in tyrosine metabolism and microsomal drug metabolism.[1]

Caution

May darken in storage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jk Psc Quick Review for Medical Officers: A Useful Manual For NEET, AIIMS and PGI Md/Ms. Entrance Exams (2018)
Dehydrofreezing of pineapple
Ramallo LA and Mascheroni RH
Journal of Food Engineering, 99(3), 269-275 (2010)
Synthesis and characterization of cycloaliphatic hydrophilic polyurethanes, modified with L-ascorbic acid, as materials for soft tissue regeneration
Kucinska-Lipka J, et al.
Materials Science & Engineering. C, Materials For Biological Applications, 75(3), 671-681 (2017)
Gemma Rush et al.
Parasitology, 146(9), 1184-1187 (2019-03-13)
Bovine trichomoniasis is a notifiable, reproductive disease of cattle caused by the parasite Tritrichomonas foetus. Culturing with modified Diamond's medium (MDM) is required to increase the low number of organisms received from a preputial sample, but is limited in application
A family of mammalian NaI[plus]I-dependent L-ascorbic acid transporters.
Tsukaguchi H, et al.
Nature, 673(1), 70-75 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service