Skip to Content
Merck
All Photos(1)

Key Documents

W530066

Sigma-Aldrich

Tocopherols

mixed, Low-α type, FCC, FG

Synonym(s):

Methyltocols

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12164502
eCl@ss:
34058016
NACRES:
NA.21
grade:
FG
Kosher
biological source:
vegetable oil
food allergen:
soybeans
Pricing and availability is not currently available.

biological source

vegetable oil

grade

FG
Kosher

reg. compliance

EU Regulation 1333/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 184.1890

refractive index

n20/D 1.501

bp

>200 °C

density

0.930 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

soybeans

Organoleptic

faint

SMILES string

O1C(CCc2c1c(c(c(c2)O)C)C)(CCCC(CCCC(CCCC(C)C)C)C)C

InChI

1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-19-26(29)23(5)24(6)27(25)30-28/h19-22,29H,8-18H2,1-7H3

InChI key

QUEDXNHFTDJVIY-UHFFFAOYSA-N

General description

Tocopherols (mixture of D-α, D-β, D-Δ, and D-γ-tocopherols) are the isomers of Vitamin E.

Application

Antioxidant used with fats and oils to delay rancidity onset

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>392.0 °F - closed cup

Flash Point(C)

> 200 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barry Halliwell et al.
The American journal of clinical nutrition, 81(1 Suppl), 268S-276S (2005-01-11)
Foods and beverages rich in phenolic compounds, especially flavonoids, have often been associated with decreased risk of developing several diseases. However, it remains unclear whether this protective effect is attributable to the phenols or to other agents in the diet.
Nazim Hussain et al.
Journal of agricultural and food chemistry, 61(26), 6137-6149 (2013-05-30)
Tocopherols are lipophilic molecules, ubiquitously synthesized in all photosynthetic organisms. Being a group of vitamin E compounds, they play an essential role in human nutrition and health. Despite their structural and functional attributes as important antioxidants in plants, it would
Chung S Yang et al.
Topics in current chemistry, 329, 21-33 (2012-07-28)
Many epidemiological studies have suggested that a low vitamin E nutritional status is associated with increased cancer risk. However, several recent large-scale human trials with high doses of α-tocopherol (α-T) have produced disappointing results. This points out the need for
William L Stone et al.
Annals of the New York Academy of Sciences, 1031, 223-233 (2005-03-09)
Colorectal cancer is the second most common cause of cancer deaths in the United States. Vitamin E (VE) and other antioxidants may help prevent colon cancer by decreasing the formation of mutagens arising from the free radical oxidation of fecal
Rosana Chirinos et al.
Food chemistry, 141(3), 1732-1739 (2013-07-23)
Fatty acids (FA), phytosterols, tocopherols, phenolic compounds, total carotenoids and hydrophilic and lipophilic ORAC antioxidant capacities were evaluated in 16 cultivars of Sacha inchi (SI) seeds with the aim to valorise them and offer more information on the functional properties

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service