Skip to Content
Merck
All Photos(1)

Key Documents

30570

Sigma-Aldrich

Decane

≥95%

Synonym(s):

n-Decane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28
Beilstein:
1696981
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
170.0-182.0 °C
174 °C (lit.)
vapor pressure:
1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

vapor density

4.9 (vs air)

Quality Level

vapor pressure

1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

Assay

≥95%

form

liquid

autoignition temp.

410 °F

expl. lim.

2.6 %

refractive index

n20/D 1.411 (lit.)
n20/D 1.412

bp

170.0-182.0 °C
174 °C (lit.)

mp

−30 °C (lit.)

density

0.730 g/mL at 20 °C
0.73 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decane is an organic solvent. Bifunctional conversion of decane over Pt/ZSM-22, Pt/ZSM-5 and Pt/USY catalysts was compared. Structure of single-phase (sodium di-2-ethylsulfosuccinate)/D2O/decane microemulsions was investigated by small-angle neutron scattering.

Application

Decane was used as solvent to investigate the self-association of cyclohexanols.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Selective conversion of decane into branched isomers: A comparison of platinum/ZSM-22, platinum/ZSM-5 and platinum/USY zeolite catalysts.
Martens JA, et al.
Applied Catalysis, 76(1), 95-116 (1991)
Self-association of cyclohexanols in inert solvents. Apparent heat capacities of cyclohexanol and substituted cyclohexanols in n-heptane and n-decane.
Trejo LM, et al.
J. Chem. Soc., Faraday, 87(11), 1739-1743 (1991)
Structure of Dense Sodium Di-2-Ethylsulfosuccinate/D 2 O/Decane Microemulsions.
Kotlarchyk M, et al.
Physical Review Letters, 53(9), 941-941 (1984)
Mahvash Karimi et al.
Colloids and surfaces. B, Biointerfaces, 95, 129-136 (2012-03-27)
Wettability alteration is considered to be one of the important mechanisms that lead to increased oil recovery during microbial enhanced oil recovery (MEOR) processes. Changes in wettability will greatly influence the petrophysical properties of the reservoir rocks and determine the
Kai Sotthewes et al.
Langmuir : the ACS journal of surfaces and colloids, 29(11), 3662-3667 (2013-02-21)
The energetics and dynamics of the various phases of decanethiolate self-assembled monolayers on Au(111) surfaces were studied with scanning tunneling microscopy. We have observed five different phases of the decanethiolate monolayer on Au(111): four ordered phases (β, δ, χ*, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service