Skip to Content
Merck
All Photos(1)

Documents

B82200

Sigma-Aldrich

4-Bromotoluene

98%

Synonym(s):

1-Methyl-4-bromobenzene, 4-Bromo-1-methylbenzene, 4-Methyl-1-bromobenzene, 4-Methylbromobenzene, 4-Tolyl bromide, p-Bromo(methyl)benzene, p-Methylbromobenzene, p-Tolyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4Br
CAS Number:
Molecular Weight:
171.03
Beilstein:
1903636
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

184 °C (lit.)

mp

26-29 °C (lit.)

density

1.39 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Br)cc1

InChI

1S/C7H7Br/c1-6-2-4-7(8)5-3-6/h2-5H,1H3

InChI key

ZBTMRBYMKUEVEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromotoluene is a p-substituted aryl bromide. 4-Bromotoluene undergoes C-O coupling reaction with 2,4-dimethylphenol catalyzed by the CuI/K2CO3/phen system. Complexes [Cu(phen)(1-(2,4-dimethylphenoxy)-4-methylbenzene)]+, {H[Cu(phen)(2,4-dimethylphenoxy)]}+ and [Cu(2,4-dimethylphenoxy)2]- were reported as intermediates during the reaction by in situ electrospray ionization mass spectrometry (ESI-MS) analysis. Suzuki coupling reaction of 4-bromotoluene and non-fluorescent phenylboronic acid (PBA) in the presence of palladium (II) acetate as a catalyst has been studied. Palladium catalyzed C-N cross-coupling reaction of 4-bromotoluene with piperidine in a microstructured continuous reactor has been described. 4-Bromotoluene has been reported to undergo Heck reaction with styrene in the presence of in situ generated palladium complexes of phosphine-functionalized N-heterocyclic carbene ligands.

Application

4-Bromotoluene was used in the synthesis of ketones by Cu/Pd-catalyzed decarboxylative cross-coupling reaction.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triaryl phosphine-functionalized N-heterocyclic carbene ligands for Heck reaction.
Wang A-E, et al.
Tetrahedron, 61(1), 259-266 (2005)
Synthesis of ketones from alpha-oxocarboxylates and aryl bromides by Cu/Pd-catalyzed decarboxylative cross-coupling.
Lukas J Goossen et al.
Angewandte Chemie (International ed. in English), 47(16), 3043-3045 (2008-03-11)
Hong-Jie Chen et al.
Dalton transactions (Cambridge, England : 2003), 43(29), 11410-11417 (2014-06-18)
The C-O coupling reaction between 2,4-dimethylphenol and 4-bromotoluene catalyzed by the CuI/K2CO3/phen system can be inhibited by the radical scavenger cumene. Complexes [Cu(i)(phen)(1-(2,4-dimethylphenoxy)-4-methylbenzene)](+) (denoted as A), {H[Cu(i)(phen)(2,4-dimethylphenoxy)]}(+) and [Cu(i)(2,4-dimethylphenoxy)2](-) (denoted as B) were observed by in situ electrospray ionization mass
Naoya Kishikawa et al.
Journal of chromatography. A, 1216(40), 6873-6876 (2009-09-01)
The fluorogenic derivatization method for aryl halide was developed for the first time. This method was based on the formation of fluorescent biphenyl structure by Suzuki coupling reaction between aryl halides and non-fluorescent phenylboronic acid (PBA). We measured the fluorescence
Successful application of microstructured continuous reactor in the palladium catalysed aromatic amination.
Mauger C, et al.
Journal of Organometallic Chemistry, 690(16), 3627-3629 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service