Skip to Content
Merck
All Photos(1)

Documents

167150

Sigma-Aldrich

1-Bromo-4-nitrobenzene

99%

Synonym(s):

1-Bromo-4-nitrobenzene, 1-Nitro-4-bromobenzene, 4-Bromo-1-nitrobenzene, 4-Nitro-1-bromobenzene, 4-Nitrobromobenzene, 4-Nitrophenyl bromide, p-Bromonitrobenzene, p-Nitrophenyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NO2
CAS Number:
Molecular Weight:
202.01
Beilstein:
636964
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

255-256 °C (lit.)

mp

124-126 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Br)cc1

InChI

1S/C6H4BrNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H

InChI key

ZDFBKZUDCQQKAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electrochemical reduction of 1-bromo-4-nitrobenzene at zinc microelectrodes in ionic liquid has been investigated by cyclic voltammetry. 1-Bromo-4-nitrobenzene undergoes palladium-catalyzed Stille cross coupling reaction with furan-2-yltributyltin using Dabco(triethylenediamine) as ligand. It reacts with p-cresol to yield 1-methyl-4-(4-nitrophenoxy)benzene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of diaryl ethers using an easy-to-prepare, air-stable, soluble copper (I) catalyst.
Gujadhur R and Venkataraman D.
Synthetic Communications, 31(18), 2865-2879 (2001)
Evgeniy G Gordeev et al.
PloS one, 13(6), e0198370-e0198370 (2018-06-08)
Additive manufacturing with fused deposition modeling (FDM) is currently optimized for a wide range of research and commercial applications. The major disadvantage of FDM-created products is their low quality and structural defects (porosity), which impose an obstacle to utilizing them
Jin-Heng Li et al.
The Journal of organic chemistry, 70(7), 2832-2834 (2005-03-25)
[reaction: see text] An efficient Pd(OAc)2/Dabco-catalyzed Stille cross-coupling reaction procedure has been developed. In the presence of Pd(OAc)2 and Dabco (triethylenediamine), various aryl halides including aryl iodides, aryl bromides, and activated aryl chlorides were coupled efficiently with organotin compounds to
Sven Ernst et al.
Physical chemistry chemical physics : PCCP, 16(10), 4478-4482 (2014-01-30)
The electrochemical reduction of 1-bromo-4-nitrobenzene (p-BrC6H4NO2) at zinc microelectrodes in the [C4mPyrr][NTf2] ionic liquid was investigated via cyclic voltammetry. The reduction was found to occur via an EC type mechanism, where p-BrC6H4NO2 is first reduced by one electron, quasi-reversibly, to
Xiao Yan et al.
Physical chemistry chemical physics : PCCP, 22(11), 6222-6230 (2020-03-05)
Unveiling the reaction mechanism is significant for developing high-performance catalysts. In this paper, a series of precisely controlled PdxM147-x (M = Cu, Pt, Au, Rh, Ru) dendrimer encapsulated nanoparticles (DENs) has been successfully synthesized. The mechanisms of PdxM147-x as catalysts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service