Skip to Content
Merck
All Photos(1)

Key Documents

B56404

Sigma-Aldrich

4′-Bromoacetophenone

98%

Synonym(s):

1-Acetyl-4-bromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4COCH3
CAS Number:
Molecular Weight:
199.04
Beilstein:
386015
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

bp

255 °C (lit.)

mp

49-51 °C (lit.)

SMILES string

CC(=O)c1ccc(Br)cc1

InChI

1S/C8H7BrO/c1-6(10)7-2-4-8(9)5-3-7/h2-5H,1H3

InChI key

WYECURVXVYPVAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4′-Bromoacetophenone is used as an organic building block in Heck and Suzuki coupling reactions. Generates phenyl radicals upon UV irradiation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gagan Chouhan et al.
Chemical communications (Cambridge, England), (45)(45), 4809-4811 (2007-11-16)
Magnetic nanoparticle-supported proline ligand was prepared and used for the CuI catalyzed Ullmann-type coupling reactions of aryl/heteroaryl bromides with various nitrogen heterocycles to form the corresponding N-aryl products in good to excellent yields; furthermore, this magnetic nanoparticle-supported proline ligand could
R Jeon et al.
Archives of pharmacal research, 24(1), 39-43 (2001-03-10)
4'-Bromoacetophenone analogs, which are able to generate monophenyl radicals capable of hydrogen atom abstraction, were investigated as possible photoinducible DNA cleaving agents. The potential of 4'-bromoacetophenone as a possible new DNA cleaver is explored. Pyrrolecarboxatmid conjugated 4'-bromoacetophenones, in particular, DNA
Mechanochemical solid-state Suzuki reactions using an in situ generated base.
Franziska Schneider et al.
ChemSusChem, 1(7), 622-625 (2008-08-15)
In vitro histamine release from basophils of asthmatic and atopic individuals in D2O.
R Tung et al.
Journal of immunology (Baltimore, Md. : 1950), 128(5), 2067-2072 (1982-05-01)
Lidiane S Araújo et al.
Marine drugs, 9(5), 889-905 (2011-06-16)
Several microorganisms were isolated from soil/sediment samples of Antarctic Peninsula. The enrichment technique using (RS)-1-(phenyl)ethanol as a carbon source allowed us to isolate 232 psychrophile/psychrotroph microorganisms. We also evaluated the enzyme activity (oxidoreductases) for enantioselective oxidation reactions, by using derivatives

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service