All Photos(2)
About This Item
Linear Formula:
CH3C6H4Br
CAS Number:
Molecular Weight:
171.03
Beilstein:
1903633
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39060138
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.552 (lit.)
bp
183.7 °C (lit.)
mp
−40 °C (lit.)
density
1.41 g/mL at 25 °C (lit.)
functional group
bromo
SMILES string
Cc1cccc(Br)c1
InChI
1S/C7H7Br/c1-6-3-2-4-7(8)5-6/h2-5H,1H3
InChI key
WJIFKOVZNJTSGO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
3-Bromotoluene is an electron-rich aryl bromide. It participates in the Heck reaction. 3-Bromotoluene undergoes palladium-catalyzed cyanation reaction in the presence of K4[Fe(CN)6] as the cyanide surrogate. It undergoes palladium-catalysed reaction with alkynyltriarylborates to yield trisubstituted alkenylboranes.
Application
3-Bromotoluene was used in the precise determination of bromine isotope ratio in organic compounds by MC-ICPMS ( Multicollector-Inductively Coupled Plasma Mass Spectrometer).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
140.0 °F
Flash Point(C)
60 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Heck reaction with an alkenylidenecyclopropane: the formation of arylallylidenecyclopropanes.
Fall Y, et al.
Tetrahedron Letters, 48(20), 3579-3581 (2007)
High precision determination of bromine isotope ratio by GC-MC-ICPMS.
Gelman F and Halicz L.
International Journal of Mass Spectrometry, 289(2), 167-169 (2010)
Naoki Ishida et al.
Chemical communications (Cambridge, England), (42)(42), 4381-4383 (2007-10-25)
The palladium-catalysed reaction of alkynyltriarylborates with aryl halides afforded trisubstituted alkenylboranes, in which two different aryl groups were installed across the carbon-carbon double bond in a cis arrangement.
Efficient cyanation of aryl bromides with K4[Fe(CN)6] catalyzed by a palladium-indolylphosphine complex.
Yeung PY, et al.
Tetrahedron Letters, 52(52), 7038-7041 (2011)
Naoyuki Yasaka et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(10), 1183-1188 (2018-10-12)
Aryl halides are a very important category of compounds that include many vital drugs and key industrial additives, such as clofibrate and bromobenzene, respectively. Due to their importance, our research group previously developed a novel fluorescence labeling approach for their
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service