925063
Silver carbonate
Technipur®, ≥99% trace metals basis
About This Item
Recommended Products
grade
reagent grade
Quality Level
Assay
≥99% trace metals basis
mp
210 °C (dec.) (lit.)
density
6.08 g/mL at 25 °C (lit.)
SMILES string
[Ag]OC(=O)O[Ag]
InChI
1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
InChI key
KQTXIZHBFFWWFW-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service