Skip to Content
Merck
All Photos(1)

Key Documents

925098

Sigma-Aldrich

Potassium tetrachloroplatinate(II)

Technipur®, ≥99.9% trace metals basis

Synonym(s):

Potassium platinum(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl4
CAS Number:
Molecular Weight:
415.09
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.23
grade:
reagent grade

grade

reagent grade

Quality Level

Assay

≥99.9% trace metals basis

density

3.38 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)Cl

InChI

1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

RVRDLMCWUILSAH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

K2PtCl4 can be used as precusor for Pt-nanoparticles. PtNPs/MoS2 composites may be used as biosensor to detect concentrations of dopamine and uric acid.

Features and Benefits

Reacts with bis(dithiolates) to form metal-bis(dithiolates) with applications in laser Q-switch materials, liquid crystals, optical CD recording media, bar code materials and superconductivity. Reacts with capping polymers to form shape-control colloidal platinum nanoparticles for catalysis and photocatalysts.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service