Skip to Content
Merck
All Photos(1)

Key Documents

363685

Sigma-Aldrich

Silver carbonate on Celite®

extent of labeling: ~50 wt. % loading

Synonym(s):

Fetizon′s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2CO3
CAS Number:
Molecular Weight:
275.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

form

powder and chunks

concentration

45-60 wt. % (by KSCN, titration)

extent of labeling

~50 wt. % loading

SMILES string

[Ag]OC(=O)O[Ag]

InChI

1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

KQTXIZHBFFWWFW-UHFFFAOYSA-L

Application

Employed in the silver ion induced preparation of α-keto acetals and α,α-dialkoxy imines. Catalyzes the generation of stereospecific unsaturated aldehydes from vinylic halides and primary amines.

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 427-427 (1994)
Tetrahedron Letters, 31, 6641-6641 (1990)
V Pacheco-Fowler et al.
The Journal of hospital infection, 57(2), 170-174 (2004-06-09)
The effect of endotracheal tubes (ETTs) impregnated with chlorhexidine (CHX) and silver carbonate (antiseptic ETTs) against Staphylococcus aureus, methicillin-resistant S. aureus (MRSA), Pseudomonas aeruginosa, Acinetobacter baumannii, and Enterobacter aerogenes [organisms associated with ventilator-associated pneumonia (VAP)], was evaluated in a laboratory
Gregory L Hamilton et al.
Journal of the American Chemical Society, 130(45), 14984-14986 (2008-10-22)
Reactions proceeding through cationic intermediates that lack a Lewis or Brønsted basic site present a challenge for traditional asymmetric catalysis based on chiral metals or organocatalysts. We present an enantioselective ring opening of tetrasubstituted meso-aziridinium ions with alcohol nucleophiles proceeding
Palladium-catalyzed decarboxylative C-H bond arylation of thiophenes.
Peng Hu et al.
Angewandte Chemie (International ed. in English), 51(1), 227-231 (2011-11-17)

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service