Skip to Content
Merck
All Photos(4)

Documents

179647

Sigma-Aldrich

Silver carbonate

99%

Synonym(s):

Disilver carbonate, Fetizon′s reagent, Silver tricarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2CO3
CAS Number:
Molecular Weight:
275.75
Beilstein:
6936654
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder and chunks

reaction suitability

reagent type: catalyst
core: silver

mp

210 °C (dec.) (lit.)

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ag]OC(=O)O[Ag]

InChI

1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

KQTXIZHBFFWWFW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Silver carbonate,prepared by precipitation or ion-exchange method can behave as a visible light sensitive semiconductor photocatalyst.

Application

Silver carbonate (Ag2CO3)promotes the regioselective benzylation of carbohydrate derivatives in high yields. Ag2CO3 can be used as a base for the Pd catalysed oxyarylation of olefins by ortho-iodophenols.It can also be used for Koenigs-Knorr glycosylation.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed oxyarylation of olefins using silver carbonate as the base. Probing the mechanism by electrospray ionization mass spectrometry.
Buarque CD, et al.
Journal of Organometallic Chemistry, 695(18), 2062-2067 (2010)
Photocatalytic behavior and photo-corrosion of visible-light-active silver carbonate/titanium dioxide.
Wang Y, et al
Materials Letters, 115, 85-88 (2014)
A novel high-efficiency visible-light sensitive Ag 2 CO 3 photocatalyst with universal photodegradation performances: Simple synthesis, reaction mechanism and first-principles study.
Dong H, et al.
Applied Catalysis. B, Environmental, 134, 46-54 (2013)
Gregory L Hamilton et al.
Journal of the American Chemical Society, 130(45), 14984-14986 (2008-10-22)
Reactions proceeding through cationic intermediates that lack a Lewis or Brønsted basic site present a challenge for traditional asymmetric catalysis based on chiral metals or organocatalysts. We present an enantioselective ring opening of tetrasubstituted meso-aziridinium ions with alcohol nucleophiles proceeding
V Pacheco-Fowler et al.
The Journal of hospital infection, 57(2), 170-174 (2004-06-09)
The effect of endotracheal tubes (ETTs) impregnated with chlorhexidine (CHX) and silver carbonate (antiseptic ETTs) against Staphylococcus aureus, methicillin-resistant S. aureus (MRSA), Pseudomonas aeruginosa, Acinetobacter baumannii, and Enterobacter aerogenes [organisms associated with ventilator-associated pneumonia (VAP)], was evaluated in a laboratory

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service