Skip to Content
Merck
All Photos(2)

Key Documents

87580

Sigma-Aldrich

Tetrakis(decyl)ammonium bromide

≥99.0% (AT)

Synonym(s):

TDAB, Tetra-decylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)9]4N(Br)
CAS Number:
Molecular Weight:
659.01
Beilstein:
5842649
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (AT)

form

powder

SMILES string

[Br-].CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC

InChI

1S/C40H84N.BrH/c1-5-9-13-17-21-25-29-33-37-41(38-34-30-26-22-18-14-10-6-2,39-35-31-27-23-19-15-11-7-3)40-36-32-28-24-20-16-12-8-4;/h5-40H2,1-4H3;1H/q+1;/p-1

InChI key

AHNISXOXSNAHBZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tetrakis(decyl)ammonium bromide is a cationic surfactant.

Tetrakis(decyl)ammonium bromide is a quaternary ammonium/phosphonium salt used as an ion-pair agent, and for the modification of bentonite.

Application

Tetrakis(decyl)ammonium bromide acts as a phase transfer agent for:
  • Platinum salts from aqueous to organic phase during the synthesis of platinum nanocrystals.
  • Gold ions from an aqueous to an organic phase during the synthesis of gold nanoparticles.
  • The alkylation of (E)‐pyridine aldoximes and (E)‐ketoximes with dihalohydrins.
It has been used as a reactant in the synthesis of ionic monomers to produce soft thermoplastic polyurethanes(TPU).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of highly uniform Pickering emulsions by mercaptocarboxylated gold nanoparticles.
Yamanaka K
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 436, 18-25 (2013)
Electron tomography and 3D molecular simulations of platinum nanocrystals.
Florea I
Nanoscale, 4(16), 5125-5131 (2012)
O-Alkylation of pyridine aldo-and ketoximes with dihalohydrins under phase-transfer conditions
Kocak A, et al.
Synthetic Communications, 37, 1155-1165 (2007)
Use of purified and modified bentonites in linear low-density polyethylene/organoclay/compatibilizer nanocomposites
Tijen S et al.
Journal of Applied Polymer Science, 124, 2430-2440 (2012)
Surface activity of water soluble amphiphilic chitin derivatives.
Desbrieres J
Polym. Bull., 39(2), 209-215 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service