
Tetrakis(decyl)ammonium bromide

Synonym(s):
Tetra-decylammonium bromide
Linear Formula:
[CH3(CH2)9]4N(Br)
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing