Skip to Content
Merck
All Photos(2)

Key Documents

87301

Sigma-Aldrich

Tetraheptylammonium bromide

≥99.0% (AT)

Synonym(s):

THPAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)6]4N(Br)
CAS Number:
Molecular Weight:
490.69
Beilstein:
3763391
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (AT)

form

flakes

mp

87-89 °C (lit.)

SMILES string

[Br-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC

InChI

1S/C28H60N.BrH/c1-5-9-13-17-21-25-29(26-22-18-14-10-6-2,27-23-19-15-11-7-3)28-24-20-16-12-8-4;/h5-28H2,1-4H3;1H/q+1;/p-1

InChI key

YQIVQBMEBZGFBY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetraheptylammonium bromide is a quaternary ammonium compound (QAC) mainly used as a phase-transfer agent. It is used as a catalyst for the Suzuki-Miyaura coupling reactions.

Application

Tetraheptylammonium bromide can be used as:
  • An alkylation reagent to synthesize benzothiazoles by using iodoanilines and sulfur.
  • A solvent in the Pd-catalyzed Suzuki and Stille coupling reactions of aryl halides.
  • A catalyst for carboxylative cyclization of propargylic amines to synthesize oxazolidinones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quaternary ammonium salt as alkylation agent in three-component reactions for the synthesis of benzothiazoles in water
Pan L, et al.
Royal Society of Chemistry Advances, 4, 27775-27779 (2014)
Trace determination of methanol in water?ethanol solution by derivatization and high-performance liquid chromatography.
Chen S
Journal of Chromatography A, 799(1-2), 93-99 (1998)
Pd nanoparticles as efficient catalysts for Suzuki and Stille coupling reactions of aryl halides in ionic liquids
Calo V, et al.
The Journal of Organic Chemistry, 70, 6040-6044 (2005)
Robust silver (I) catalyst for the carboxylative cyclization of propargylic alcohols with carbon dioxide under ambient conditions
Song QW and He LN
Advanced Synthesis & Catalysis, 358, 1251-1258 (2016)
Carbon-carbon cross coupling reactions in ionic liquids catalysed by palladium metal nanoparticles
Prechtl M, et al.
Molecules (Basel), 15, 3441-3461 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service