Skip to Content
Merck
All Photos(2)

Documents

80980

Sigma-Aldrich

Platinum on activated charcoal

greener alternative

hydrogenation catalyst

Synonym(s):

Pt/C

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

hydrogenation catalyst

form

powder

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

matrix

activated charcoal support

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Platinum on activated carbon (Pt/C) can be used as a catalyst for a variety of applications such as:
  • hydrogenation of D-fructose and D-glucose
  • hydrosilylation reaction for the production of molecules with silicon-carbon bonds
  • direct alcohol fuel cells (DAFCs)

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of thermoreactive polysiloxanes with cyclic fragments in the side chain
Mukbaniani O, et al.
Journal of Applied Polymer Science, 114(2), 892-900 (2009)
Preparation of platinum on activated carbon
Van Dam HE, et al.
J. Catal., 131(2), 335-349 (1991)
Platinum catalysts supported on activated carbons: I. Preparation and characterization
Rodriguez-Reinoso F, et al.
J. Catal., 99(1), 171-183 (1986)
Hydrogenation of d-fructose over activated charcoal supported platinum catalyst
Ahmed MJ and Kadhum AAH
Journal of the Taiwan Institute of Chemical Engineers, 42(1), 114-119 (2011)
Melillo, D. G.; Cvetovich, R. J. et al.
The Journal of Organic Chemistry, 51, 1498-1498 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service