Skip to Content
Merck
All Photos(3)

Key Documents

237558

Sigma-Aldrich

Platinum on carbon

greener alternative

extent of labeling: 3 wt. % loading, matrix activated carbon support

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

3 wt. % loading

matrix

activated carbon support

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Platinum on carbon (Pt/C) can be used as a catalyst for a variety of applications, such as:
  • hydrosilylation reaction for the production of molecules with silicon-carbon bonds
  • direct alcohol fuel cells (DAFCs)
  • an electrocatalyst in hydrogen evolution reaction (HER)
  • formation of nanocatalyst for polymer electrolyte fuel cells
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platinum supported on mesoporous carbon as cathode catalyst for direct methanol fuel cells
Bruno MM, et al.
Journal of Power Sources, 278, 458-463 (2015)
Hydrosilylation of alkynes catalyzed by platinum on carbon.
Chauhan M, et al.
Journal of Organometallic Chemistry, 645(1), 1-13 (2002)
Synthesis and physical/electrochemical characterization of Pt/C nanocatalyst for polymer electrolyte fuel cells
Prabhuram J, et al.
Journal of Power Sources, 134(1), 1-6 (2004)
Highly efficient hydrogen evolution from seawater by a low-cost and stable CoMoP@ C electrocatalyst superior to Pt/C
Ma Y, et al.
Energy & Environmental Science, 10(3), 788-798 (2017)
Ethanol oxidation reactions using SnO2@ Pt/C as an electrocatalyst
Silva JCM, et al.
Applied Catalysis. B, Environmental, 99(1-2), 265-271 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service