Skip to Content
Merck
All Photos(3)

Documents

238058

Sigma-Aldrich

Ruthenium(IV) oxide

99.9% trace metals basis

Synonym(s):

Ruthenium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RuO2
CAS Number:
Molecular Weight:
133.07
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: ruthenium

density

6.97 g/mL at 25 °C (lit.)

SMILES string

O=[Ru]=O

InChI

1S/2O.Ru

InChI key

WOCIAKWEIIZHES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ruthenium oxide is an inorganic compound widelyused as electrode material due to its high thermodynamic and chemicalstability. It is also used in supercapacitor applications because of its widepotential window for reversible redox reactions and a long-life cycle.

Application

Ruthenium(IV)oxide can be used:

  • As a catalyst for hydrogen evolution reaction.
  • In the preparation of thin-film supercapacitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meixuan Li et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(2), 1901833-1901833 (2020-01-30)
Developing high-performance, low-cost, and robust bifunctional electrocatalysts for overall water splitting is extremely indispensable and challenging. It is a promising strategy to couple highly active precious metals with transition metals as efficient electrocatalysts, which can not only effectively reduce the
Huayu Qian et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(50), 18259-18264 (2016-11-01)
Cobalt sulfide/sulfur doped carbon composites (Co9 S8 /S-C) were synthesized by calcining a rationally designed sulfur-containing cobalt coordination complex in an inert atmosphere. From the detailed transmission electron microscopy (TEM) and X-ray photoelectron spectroscopy (XPS) analyses, the electrocatalytically active Co9
B Sachin Kumar et al.
Dalton transactions (Cambridge, England : 2003), 48(33), 12684-12698 (2019-08-07)
Producing pure H2 and O2 to sustain the renewable energy sources with minimal environmental damage is a key objective of photo/electrochemical water-splitting research. Metallic Ni-based electrocatalysts are expensive and eco-hazardous. This has rendered the replacement or reduction of Ni content
S Raghu et al.
Journal of hazardous materials, 139(2), 381-390 (2006-09-30)
The paper presents the results of an efficient electrochemical treatment of Procion Black 5B--a pilot plant study. Experiments were conducted at different current densities and selected electrolyte medium using Ti/RuO2 as anode, stainless-steel as cathode in a cylindrical flow reactor.
Naiqiang Yan et al.
Environmental science & technology, 45(13), 5725-5730 (2011-06-15)
Catalytic conversion of elemental mercury (Hg(0)) to its oxidized form has been considered as an effective way to enhance mercury removal from coal-fired power plants. In order to make good use of the existing selective catalytic reduction of NO(x) (SCR)

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service