Skip to Content
Merck
All Photos(3)

Key Documents

520691

Sigma-Aldrich

Platinum on silica

greener alternative

extent of labeling: 1 wt. % loading, granular, dry

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

dry
granular

reaction suitability

reagent type: catalyst
core: platinum

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

1 wt. % loading

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Application

Platinum on silica (Pt/SiO2) can be used for a variety of applications such as:
  • a substrate for the preparation of zinc oxide (ZnO) nanopillar arrays
  • an electrocatalyst for methanol oxidation
  • in the formation of nanospheres for catalytic hydrogen production

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoelectrochemical hydrogen production of TiO2 passivated Pt/Si-nanowire composite photocathode
Li S, et al.
ACS Applied Materials & Interfaces, 7(33), 18560-18565 (2015)
Spatially separated ZnO nanopillar arrays on Pt/Si substrates prepared by electrochemical deposition
Lee S, et al.
The Journal of Physical Chemistry C, 111(32), 11793-11801 (2007)
Core-shell nanospheres Pt@ SiO2 for catalytic hydrogen production
Hu Y, et al.
Applied Surface Science, 341, 185-189 (2015)
Hydrogen production by glycerol steam reforming with Pt/SiO2 and Ni/SiO2 catalysts
Pompeo F, et al.
Catalysis Today, 172(1), 183-188 (2011)
Solvothermal synthesis of Pt-SiO2/graphene nanocomposites as efficient electrocatalyst for methanol oxidation
Vu THT, et al.
Electrochimica Acta, 161, 335-342 (2015)

Articles

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service