Skip to Content
Merck
All Photos(1)

Key Documents

676403

Sigma-Aldrich

(R,R)-(–)-2,3-Bis(tert-butylmethylphosphino)quinoxaline

≥95%

Synonym(s):

(R) QuinoxP®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2P2
CAS Number:
Molecular Weight:
334.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95%

form

solid

mp

100-104 °C

functional group

phosphine

SMILES string

CP(c1nc2ccccc2nc1P(C)C(C)(C)C)C(C)(C)C

InChI

1S/C18H28N2P2/c1-17(2,3)21(7)15-16(22(8)18(4,5)6)20-14-12-10-9-11-13(14)19-15/h9-12H,1-8H3/t21-,22-/m0/s1

InChI key

DRZBLHZZDMCPGX-VXKWHMMOSA-N

General description

(R,R)-(-)-2,3-Bis(tert-butylmethylphosphino)quinoxaline is an air-stable C2-symmetric P-stereogenic phosphine ligand.[1]

Application

Efficient ligand exhibiting high levels of enantiocontrol in synthetic transformations ranging from metal-catalyzed 1,4-addition of arylboronic acids to alkylative ring opening to asymmetric hydrogenation.[2]

Features and Benefits

Advantages of the QuinoxP* Ligands:
  • It is not oxidized nor epimerized at ambient conditions in air
  • Enantioselectivities are outstanding for various reaction paradigms
  • Hydrogenations proceed under mild reaction conditions
  • Low catalyst loadings yield high TONs

Legal Information

QuinoxP is a registered trademark of Nippon Chemical Industry Co., Ltd.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Copper-Catalyzed Propargylic Substitution Reaction of Propargylic Acetates with Enamines.
Fang P and Hou XL.
Organic Letters, 11.20, 4612-4615 (2009)
Highly enantioselective hydrosilylation of simple ketones catalyzed by rhodium complexes of P-chiral diphosphine ligands bearing tert-butylmethylphosphino groups.
Imamoto T, et al.
Tetrahedron Asymmetry, 17(4), 560-565 (2006)
A rational pre-catalyst design for bis-phosphine mono-oxide palladium catalyzed reactions.
Ji Y, et al.
Chemical Science (2017)
Optimization of the Preparation of (R)-3, 5-Bis (trifluoromethyl)-?-methyl-N-methylbenzylamine l-(?)-Malic Acid Salt through Classical Resolution.
Brandel CM, et al.
Organic Process Research & Development, 19(12), 1954-1965 (2015)
Rhodium?Catalyzed Dynamic Kinetic Asymmetric Allylation of Phenols and 2?Hydroxypyridines.
Li C and Breit B
European Journal of Chemistry, 22(41), 14655-14663 (2016)

Articles

QuinoxP*: Air-Stable and Highly Efficient and Productive Chiral Ligands

QuinoxP*: Air-Stable and Highly Efficient and Productive Chiral Ligands

QuinoxP*: Air-Stable and Highly Efficient and Productive Chiral Ligands

QuinoxP*: Air-Stable and Highly Efficient and Productive Chiral Ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service