Skip to Content
Merck
All Photos(1)

Key Documents

692484

Sigma-Aldrich

(R)-DTBM-SEGPHOS®

Synonym(s):

(R)-(−)-5,5′-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′-bi-1,3-benzodioxole, [(4R)-(4,4′-bi-1,3-benzodioxole)-5,5′-diyl]bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H100O8P2
CAS Number:
Molecular Weight:
1179.53
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder

functional group

phosphine

SMILES string

P(c8cc(c(c(c8)C(C)(C)C)OC)C(C)(C)C)(c7cc(c(c(c7)C(C)(C)C)OC)C(C)(C)C)c1c(c6c(cc1)OCO6)c2c3c(ccc2P(c5cc(c(c(c5)C(C)(C)C)OC)C(C)(C)C)c4cc(c(c(c4)C(C)(C)C)OC)C(C)(C)C)OCO3

InChI

1S/C74H100O8P2/c1-67(2,3)47-33-43(34-48(61(47)75-25)68(4,5)6)83(44-35-49(69(7,8)9)62(76-26)50(36-44)70(10,11)12)57-31-29-55-65(81-41-79-55)59(57)60-58(32-30-56-66(60)82-42-80-56)84(45-37-51(71(13,14)15)63(77-27)52(38-45)72(16,17)18)46-39-53(73(19,20)21)64(78-28)54(40-46)74(22,23)24/h29-40H,41-42H2,1-28H3

InChI key

ZNORAFJUESSLTM-UHFFFAOYSA-N

Application

Catalyst involved in:
  • [3,3]-Sigmatropic rearrangements using cyclopropane probes
  • Asymmetric intramolecular hydroacylation of ketoaldehydes

Reactant involved in:
  • The synthesis of gold-diphosphine complexes for use as catalysts
  • Cycloaddition of allenenes to yield alkylidenecyclobutanes

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service