Skip to Content
Merck
All Photos(1)

Key Documents

700754

Sigma-Aldrich

(R)-SDP

Synonym(s):

(R)-(+)-7,7′-Bis(diphenylphosphino)-2,2′,3,3′-tetrahydro-1,1′-spirobiindene, (R)-7,7′-Bis(diphenylphosphino)-1,1′-spirobiindane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H34P2
CAS Number:
Molecular Weight:
588.66
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

Quality Level

optical activity

[α]22/D +184°, c = 1 in chloroform

reaction suitability

reagent type: ligand

mp

198-203 °C

functional group

phosphine

storage temp.

−20°C

SMILES string

P(c8ccccc8)(c7ccccc7)c1c2c(ccc1)CC[C@@]32CCc4c3c(ccc4)P(c6ccccc6)c5ccccc5

InChI

1S/C41H34P2/c1-5-17-33(18-6-1)42(34-19-7-2-8-20-34)37-25-13-15-31-27-29-41(39(31)37)30-28-32-16-14-26-38(40(32)41)43(35-21-9-3-10-22-35)36-23-11-4-12-24-36/h1-26H,27-30H2

InChI key

DKZNVWNOOLQCMJ-UHFFFAOYSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service