Skip to Content
Merck
All Photos(1)

Key Documents

693065

Sigma-Aldrich

(R)-BINAP

Synonym(s):

(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene, (R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl, (R)-(+)-(1,1′-Binaphthalene-2,2′-diyl)bis(diphenylphosphine), (R)-(+)-BINAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PC10H6-]2
CAS Number:
Molecular Weight:
622.67
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

crystals

Quality Level

optical activity

[α]20/D +222°, c = 0.5% in benzene

mp

239-241 °C (lit.)

functional group

phosphine

SMILES string

c1ccc(cc1)P(c2ccccc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccccc7)c8ccccc8

InChI

1S/C44H32P2/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-32H

InChI key

MUALRAIOVNYAIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-BINAP reacts with allylpalladium(II) chloride dimer to form a BINAP-palladium catalyst, which can catalyze the asymmetric allylation of 1,3-diketones to form chiral 2,2-dialkyl-1,3-diketones. It may also be used in the preparation of chirally stabilized rhodium nanoparticles, which can be used as a catalyst for the asymmetric hydroformylation of styrene and vinyl acetate.

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric hydrogenation
Noyori, Ryoji
Acta Chemica Scandinavica, 50, 380-390 (1996)
RuHCl (diphosphine)(diamine): Catalyst Precursors for the Stereoselective Hydrogenation of Ketones and Imines1
Abdur-Rashid, Kamaluddin, Alan J. Lough, and Robert H. Morris
Organometallics, 20.6, 1047-1049 (2001)
Synthesis and resolution of the biaryl-like diphosphine (S)-Me2-CATPHOS, preparation of a derived rhodium precatalyst and applications in asymmetric hydrogenation
Doherty, Simon, and Catherine H. Smyth
Nature Protocols, 7.10, 1884-1896 (2012)
Asymmetric hydroformylation of olefins catalyzed by rhodium nanoparticles chirally stabilized with (R)-BINAP ligand.
Han D, et al.
J. Mol. Catal. A: Chem., 283(1), 15-22 (2008)
Bis [(R)?(+)?binap] rhodium (I) Perchlorate, a Highly Efficient Catalyst for the Asymmetric Isomerization of Allylamines
Tani, Kazuhide, et al.
Angewandte Chemie (International Edition in English), 24.3, 217-219 (1985)

Articles

We present an article concerning BINAP/SEGPHOS® Ligands and Complexes.

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service