Skip to Content
Merck
All Photos(2)

Key Documents

M9005

Sigma-Aldrich

3-Methyl-L-histidine

≥98% (TLC)

Synonym(s):

π-Methyl-L-histidine, 1-Methyl-L-histidine (archaic), 3-(1-Methylimidazol-5-yl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3O2
CAS Number:
Molecular Weight:
169.18
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

3-Methyl-L-histidine,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white to off-white

mp

254 °C

application(s)

detection

SMILES string

Cn1cncc1C[C@H](N)C(O)=O

InChI

1S/C7H11N3O2/c1-10-4-9-3-5(10)2-6(8)7(11)12/h3-4,6H,2,8H2,1H3,(H,11,12)/t6-/m0/s1

InChI key

JDHILDINMRGULE-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Engineering mutually orthogonal PylRS/tRNA pairs for dual encoding of functional histidine analogues.: Exploring innovative genetic engineering techniques, this research advances the use of histidine analogues like 3-Methyl-L-histidine for producing novel proteins with enhanced functionalities, offering vast potential in therapeutic and industrial applications (Taylor et al., 2023).

  • Improving the enzymatic activity of l-amino acid α-ligase for imidazole dipeptide production by site-directed mutagenesis.: This investigation enhances the enzymatic synthesis of imidazole dipeptides, crucial for understanding the role of modifications like those seen in 3-Methyl-L-histidine and its impact on biological activities (Kino et al., 2023).

Biochem/physiol Actions

3-Methyl-L-histidine is a non-proteinogenic amino acid and an index of muscle breakdown.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Lee et al.
Analytical chemistry, 82(7), 2959-2968 (2010-03-03)
Despite several decades of active research, the success of large-scale clinical trials involving antioxidants remains equivocal given the complex biological interactions of reactive oxygen/nitrogen species in human health. Herein, we outline a differential metabolomics strategy by capillary electrophoresis-electrospray ionization-mass spectrometry
S G A van der Drift et al.
Journal of dairy science, 95(9), 4911-4920 (2012-08-25)
The objective of this study was to obtain information on variation between dairy cows in muscle and fat tissue mobilization around parturition and to study the association between protein and fat mobilization and serum β-hydroxybutyrate (BHBA) concentrations (hyperketonemia) in this
M N Goodman
The American journal of physiology, 260(5 Pt 1), E727-E730 (1991-05-01)
The metabolic response to infection includes loss of lean tissue and increased nitrogen excretion. The loss of muscle tissue during infection results in large part from accelerated skeletal muscle protein breakdown. Recent studies suggest that macrophage-derived products secreted during infection
E Roth et al.
Clinical chemistry, 31(8), 1305-1309 (1985-08-01)
We measured amino acid concentrations in plasma and skeletal muscle of three groups of patients with acute hemorrhagic pancreatitis: (a) patients without secondary organ lesions, (b) patients also suffering from kidney damage, and (c) patients in whom the pancreatitis was
Rokhsareh F Vesali et al.
Clinical science (London, England : 1979), 118(6), 421-427 (2009-09-16)
The human endotoxin model has been used to study the early phase of sepsis. The aim of the present study was to assess leg muscle protein kinetics after an endotoxin challenge given to healthy human volunteers. Six healthy male subjects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service