Skip to Content
Merck
All Photos(3)

Key Documents

452483

Sigma-Aldrich

1-Methyl-D-tryptophan

95%, for peptide synthesis

Synonym(s):

(2R)-2-Amino-3-(1-methyl-1H-indol-3-yl)propanoic acid, D-(+)-1-Methyltryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

1-Methyl-D-tryptophan, 95%

Assay

95%

form

lumps and powder

optical activity

[α]22/D +12.4°, c = 2 in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

242-245 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cn1cc(C[C@@H](N)C(O)=O)c2ccccc12

InChI

1S/C12H14N2O2/c1-14-7-8(6-10(13)12(15)16)9-4-2-3-5-11(9)14/h2-5,7,10H,6,13H2,1H3,(H,15,16)/t10-/m1/s1

InChI key

ZADWXFSZEAPBJS-SNVBAGLBSA-N

Application

Used to prepare a number of natural products including macroline alkaloids using the Pictet-Spengler reaction of tryptophan esters with aldehydes also used in the synthesis of 1-substituted indolactam precursors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irie, K. et al.
Tetrahedron, 51, 6255-6255 (1995)
Zhidan Tu et al.
Stem cells and development, 19(11), 1803-1809 (2010-02-19)
Mesenchymal stem cells (MSCs) possess potent and broad immunosuppressive capabilities, and have shown promise in clinical trials treating many inflammatory diseases. Previous studies have found that MSCs inhibit dendritic cell, T-cell, and B-cell activities in the adaptive immunity; however, whether
Franco Carranza et al.
PloS one, 7(7), e40356-e40356 (2012-08-01)
Dendritic cells (DC) have the potential to control the outcome of autoimmunity by modulating the immune response. In this study, we tested the ability of Fasciola hepatica total extract (TE) to induce tolerogenic properties in CpG-ODN (CpG) maturated DC, to
Longhao Sun et al.
Journal of personalized medicine, 12(12) (2022-12-24)
Pancreatic ductal adenocarcinoma (PDAC) represents one of the deadliest malignancies. Elevated regulatory T cell (Treg) infiltration has a potent immunosuppressive function in tumor biology, which contributes to low survival in PDAC. Nonetheless, the crosstalk between malignant cells and tumor-infiltrating Tregs
Valeria Caffi et al.
Frontiers in veterinary science, 7, 318-318 (2020-07-14)
Mesenchymal stem/stromal cells (MSCs) are increasingly explored for the treatment of degenerative and inflammatory diseases in human and veterinary medicine. One of the key characteristics of MSCs is that they modulate inflammation mainly through the secretion of soluble mediators. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service