Skip to Content
Merck
All Photos(4)

Key Documents

244511

Sigma-Aldrich

Toluene

anhydrous, 99.8%

Synonym(s):

1-Methylbenzene, Methacide, Toluol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39011102
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

99.8%

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

impurities

<0.001% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

color

colorless

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene, a flammable liquid with a pungent odor, is widely employed as an organic solvent. It is also used as a precursor for synthesizing benzene.[1]

Application

Toluene undergoes alkylation in the presence of modified ZSM (Zeolite Socony Mobil)-5-class zeolite catalysts to form p-xylene with high selectivity.[2] When doped on graphene, it acts as an electron donor leading to alteration in graphene electrical properties due to electrochemical interaction.[3]

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

F Nelson Nunalee et al.
Analytical chemistry, 78(4), 1158-1166 (2006-02-16)
Spreading of liquids and soft solids on a rigid surface in a surrounding liquid medium is investigated by utilizing the lateral sensitivity of the quartz crystal microbalance (QCM). While the QCM has been used extensively to study systems with spatial
von Burg R
Journal of Applied Toxicology, 13(6) (1993)
The effect of a fullerene water suspension on the growth, cell viability, and membrane integrity of Escherichia coli B23.
Aquino A, et al.
Journal of Experimental Microbiology and Immunology, 14, 13-20 (2010)
Selective alkylation of toluene with methanol to produce para-xylene.
Kaeding WW, et al.
J. Catal., 67(1), 159-174 (1981)
Electrochemical doping of graphene with toluene.
Kaverzin AA, et al.
Carbon, 49(12), 3829-3834 (2011)

Articles

JosiPhos CyPF-tBu and palladium give catalyst for alkoxylation of activated heteroaryl halides with primary, secondary, and tertiary alcohols

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service