Skip to Content
Merck
All Photos(1)

Documents

P7888

Sigma-Aldrich

DL-Propargylglycine

cystathionine γ-lyase inhibitor

Synonym(s):

2-Amino-4-pentynoic acid, PAG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO2
CAS Number:
Molecular Weight:
113.11
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

NC(CC#C)C(O)=O

InChI

1S/C5H7NO2/c1-2-3-4(6)5(7)8/h1,4H,3,6H2,(H,7,8)

InChI key

DGYHPLMPMRKMPD-UHFFFAOYSA-N

Application


  • Metabolic Labeling Strategy Boosted Antibacterial Efficiency for Photothermal and Photodynamic Synergistic Bacteria-Infected Wound Therapy.: This study demonstrates the enhanced antibacterial efficiency of a metabolic labeling strategy using ᴅʟ-Propargylglycine in photothermal and photodynamic therapy for treating bacteria-infected wounds (Li et al., 2022).

  • Metabolic Labeling of Peptidoglycan with NIR-II Dye Enables In Vivo Imaging of Gut Microbiota.: This research utilizes ᴅʟ-Propargylglycine for metabolic labeling, allowing near-infrared II imaging of gut microbiota in vivo, providing new insights into microbiome studies (Wang et al., 2020).

  • Hydrogen sulfide upregulated mRNA expressions of sodium bicarbonate cotransporter1, trefoil factor1 and trefoil factor2 in gastric mucosa in rats.: Investigates the role of ᴅʟ-Propargylglycine, in upregulating specific mRNA expressions in gastric mucosa, with implications for gastrointestinal research (Cheraghi et al., 2016).

Biochem/physiol Actions

DL-Propargylglycine (PAG) is an irreversible inhibitor of the enzyme cystathionine γ-lyase (GCL), a key enzyme involved in glutathione synthesis and the metabolic transsulfuration pathway which regulates homocysteine concentration and mediates cysteine synthesis. Treatment of animal disease models with PAG was shown to alleviate symptoms of diseases that impair the formation or action of hydrogen sulfide (H2S).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lambert Nzungize et al.
Journal of drug targeting, 27(9), 1004-1016 (2019-02-08)
Tuberculosis, especially multidrug resistant cases, remains an enormous public health threat. Mycobacterium tuberculosis metC (Rv3340) an enzyme involved in methionine biosynthesis was identified and characterised for antimicrobial susceptibility. We reported that the overexpression of Rv3340 in Mycobacterium smegmatis (Ms_Rv3340) produces
Karen P Maruska et al.
The Journal of comparative neurology, 525(3), 610-638 (2016-08-11)
Neural communication depends on release and reception of different neurotransmitters within complex circuits that ultimately mediate basic biological functions. We mapped the distribution of glutamatergic, GABAergic, and cholinergic neurons in the brain of the African cichlid fish Astatotilapia burtoni using
Patrick Grabher et al.
NeuroImage. Clinical, 15, 494-501 (2017-06-27)
Brainstem networks are pivotal in sensory and motor function and in recovery following experimental spinal cord injury (SCI). To quantify neurodegeneration and its relation to clinical impairment in major brainstem pathways and nuclei in traumatic SCI. Quantitative MRI data of
Csaba Szabó
Nature reviews. Drug discovery, 6(11), 917-935 (2007-10-20)
Hydrogen sulphide (H2S) is increasingly being recognized as an important signalling molecule in the cardiovascular and nervous systems. The production of H2S from L-cysteine is catalysed primarily by two enzymes, cystathionine gamma-lyase and cystathionine beta-synthase. Evidence is accumulating to demonstrate
Hicham Chahtane et al.
eLife, 7 (2018-08-29)
To anticipate potential seedling damage, plants block seed germination under unfavorable conditions. Previous studies investigated how seed germination is controlled in response to abiotic stresses through gibberellic and abscisic acid signaling. However, little is known about whether seeds respond to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service