K8625
L-Kynurenine
≥98% (HPLC)
Synonym(s):
β-Anthraniloyl-L-alanine, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H12N2O3
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
L-Kynurenine, ≥98% (HPLC)
Assay
≥98% (HPLC)
form
powder
mp
219 °C
application(s)
detection
SMILES string
N[C@@H](CC(=O)c1ccccc1N)C(O)=O
InChI
1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)/t8-/m0/s1
InChI key
YGPSJZOEDVAXAB-QMMMGPOBSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
L-Kynurenine has been used as a standard for indoleamine-2,3-dioxygenase (IDO) assay. It has also been used as a standard to extract and quantify kynurenine from cultured cells and media.
Biochem/physiol Actions
Key intermediate in the breakdown pathway of tryptophan.
L-Kynurenine is a key intermediate in the breakdown of L-tryptophan and the formation of nicotinamide adenine dinucleotide (NAD+) via the kynurenine pathway. It is involved in a variety of neurological processes and diseases. L-Kynurenine is a substrate for kynureninase/kynurenine hydrolase; kynurenine 3-monooxygenase and kynurenine-oxoglutarate transaminase.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kynurenine Pathway Metabolites in Humans: Disease and Healthy States
Yiquan Chen
International Journal of Tryptophan Research : IJTR, 2, 1-19 (2009)
The Involvement of Neuroinflammation and Kynurenine Pathway in Parkinson's Disease
Anna Zinger
Parkinson's Disease (2011)
Essential and Non-Essential Metals Metabolites with Antibiotic Activity Pharmacology of Benzodiazepines Interferon Gamma Research (2012)
Evaluation of radiofluorinated carboximidamides as potential IDO-targeted PET tracers for cancer imaging
Xuan Huang
Oncotarget, 8(29), 46900-46914 (2017)
Extraction and Quantification of Tryptophan and Kynurenine from Cultured Cells and Media Using a High Performance Liquid Chromatography (HPLC) System Equipped with an Ultra-Sensitive Diode Array Detector
Jeffrey Kim
Bio-protocol, 6(7), e1781-e1781 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service