Skip to Content
Merck
All Photos(3)

Documents

C11804

Sigma-Aldrich

4-Sulfamoylbenzoic acid

97%

Synonym(s):

4-Carboxybenzenesulfonamide, Benzoic acid 4-sulfamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CC6H4SO2NH2
CAS Number:
Molecular Weight:
201.20
Beilstein:
1875393
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

285-295 °C (lit.)

functional group

carboxylic acid
sulfonamide

SMILES string

O=S(C(C=C1)=CC=C1C(O)=O)(N)=O

InChI

1S/C7H7NO4S/c8-13(11,12)6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)(H2,8,11,12)

InChI key

UCAGLBKTLXCODC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

4-Sulfamoylbenzoic acid also known as 4-carboxybenzenesulfonamide, is a sulfonamide derivative of benzoic acid. It is a crosslinking reagent used widely in organic synthesis.

Application

4-Sulfamoylbenzoic acid is used to synthesize 4-sulfamoylbenzoyl chloride and isocyanate derivatives. Additionally, it can be used in the esterification of alcohols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel synthesized SLC-0111 thiazole and thiadiazole analogues: Determination of their carbonic anhydrase inhibitory activity and molecular modeling studies
MF Abo-Ashour
Bioorganic Chemistry, 87, 794-802 (2019)
Furan-based inhibitors of pyruvate dehydrogenase: SAR study, biochemical evaluation and computational analysis
AHY Chan
Organic & Biomolecular Chemistry, 21, 1755-1763 (2023)
Jean-Yves Winum et al.
Journal of medicinal chemistry, 48(6), 2121-2125 (2005-03-18)
Targeting proteins overexpressed in hypoxic tumors is as an important means of controlling cancer disease. One such protein is the carbonic anhydrase (CA) isoenzyme IX, which in some types of tumors is overexpressed 150-200-fold. We report here a series of
Daniel J Orton et al.
Analytical chemistry, 90(1), 737-744 (2017-11-22)
To better understand disease conditions and environmental perturbations, multiomic studies combining proteomic, lipidomic, and metabolomic analyses are vastly increasing in popularity. In a multiomic study, a single sample is typically extracted in multiple ways, and various analyses are performed using
Microwave assisted synthesis of novel hybrid tacrine-sulfonamide derivatives and investigation of their antioxidant and anticholinesterase activities
R Ulus
Bioorganic Chemistry, 245-255 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service