Skip to Content
Merck
All Photos(2)

Documents

369314

Sigma-Aldrich

Oleyl alcohol

technical grade, 85%

Synonym(s):

Z-9-Dodecen-1-ol, cis-9-Octadecen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2OH
CAS Number:
Molecular Weight:
268.48
Beilstein:
1723962
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

form

liquid

refractive index

n20/D 1.46 (lit.)

bp

207 °C/13 mmHg (lit.)

mp

0-5.0 °C (lit.)

density

0.849 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC\C=C/CCCCCCCCO

InChI

1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3/b10-9-

InChI key

ALSTYHKOOCGGFT-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oleyl alcohol has been reported as an effective solvent for oil/water-type tacrolimus (FK506) cream formulation, useful for atopic dermatitis treatment. It was reported as diluting agent for Alamine 336. Palm oil esters derived from oleyl alcohol, have potential applications in the cosmetic and pharmaceutical industries due to the excellent wetting behavior of the esters without the oily feel.

Application

Oleyl alcohol may be employed as solvent during butanol production from corn stover hydrolysates (CSH) with in-situ liquid-liquid extraction.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sook Han Ng et al.
Journal of nanobiotechnology, 11, 27-27 (2013-09-26)
Palm oil esters (POEs) are esters derived from palm oil and oleyl alcohol have great potential in the cosmetic and pharmaceutical industries due to the excellent wetting behavior of the esters without the oily feel. The role of oil-in-water nanoemulsions
George M Teke et al.
Biotechnology and bioengineering, 118(1), 58-71 (2020-09-03)
Extractive fermentation (or in situ product removal (ISPR)) is an operational method used to combat product inhibition in fermentations. To achieve ISPR, different separation techniques, modes of operation and physical reactor configurations have been proposed. However, the relative paucity of
Müge Gökdere et al.
Preparative biochemistry & biotechnology, 44(1), 80-89 (2013-10-15)
Gibberelic acid fermentation using extractive methods was carried out in the presence of corn oil and Alamine 336. Gibberella fujikuroi fungus (NRRL 2278) was used to produce gibberellic acid. Oleyl alcohol was a diluting agent for Alamine 336. The effects
Masayuki Yamanaka et al.
International journal of pharmaceutics, 464(1-2), 19-26 (2014-01-25)
We developed an oil/water-type tacrolimus (FK506) cream formulation as an alternative to Protopic ointment for atopic dermatitis treatment. We determined the effects of solvents used in topical preparations on FK506 solubility and stability, and evaluated FK506 transdermal absorption into rat
Fengqin Wang et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 29(10), 1515-1526 (2014-01-18)
Butanol production from corn stover hydrolysates (CSH) with in-situ liquid-liquid extraction was studied to enhance the production and reduce the fermentation cost. Oleyl alcohol was selected as the suitable solvent and added at the initial fermentation time with the ratio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service