Skip to Content
Merck
All Photos(1)

Key Documents

8.07680

Sigma-Aldrich

Stearyl alcohol

for synthesis

Synonym(s):

Stearyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38O
CAS Number:
Molecular Weight:
270.49
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-017-6
NACRES:
NA.22
Assay:
≥96.0% (GC)
bp:
330-360 °C/1013 hPa
vapor pressure:
<1 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

form

scales

autoignition temp.

230 °C (DIN 51794)

potency

>2000 mg/kg LD50, oral (Rat)

bp

330-360 °C/1013 hPa

mp

55-60 °C

transition temp

flash point 195 °C

density

0.805-0.815 g/cm3 at 60 °C

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

OCCCCCCCCCCCCCCCCCC

InChI

1S/C18H38O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

GLDOVTGHNKAZLK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 56 °C
Melting range (upper value): ≤ 59 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

383.0 °F

Flash Point(C)

195 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service